Skip to Content
Merck
All Photos(2)

Key Documents

31464

Supelco

Anilazine

PESTANAL®, analytical standard

Synonym(s):

6-(2-Chloroanilino)-2,4-dichloro-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5Cl3N4
CAS Number:
Molecular Weight:
275.52
Beilstein:
223133
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

Clc1nc(Cl)nc(Nc2ccccc2Cl)n1

InChI

1S/C9H5Cl3N4/c10-5-3-1-2-4-6(5)13-9-15-7(11)14-8(12)16-9/h1-4H,(H,13,14,15,16)

InChI key

IMHBYKMAHXWHRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D E Breithaupt et al.
Chemosphere, 41(9), 1401-1406 (2000-11-01)
Photoreactions, initiated by sunlight irradiation, between organochlorine pesticides and olefinic compounds of plant cuticles have been postulated. Concerning the formation of bound residues, which so far have not been detectable by common analytical techniques, photoaddition reactions are of main interest.
Dyrene dermatitis.
S H Schuman et al.
Lancet (London, England), 2(8206), 1252-1252 (1980-12-06)
J Liebich et al.
Journal of agricultural and food chemistry, 47(9), 3905-3910 (1999-12-20)
Humic substance fractions obtained from a degraded loess soil taken from a long-term lysimeter experiment with the fungicide anilazine were incubated in aerated liquid cultures together with native soil microorganisms. Biomineralization, remobilization of [U-phenyl-(14)C]anilazine, respectively, its metabolites, and changes of
F C Garg et al.
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten und Hygiene. Zweite naturwissenschaftliche Abteilung: Mikrobiologie der Landwirtschaft der Technologie und des Umweltschutzes, 136(6), 505-508 (1981-01-01)
The effect of anilazine on growth, respiration and enzyme activity of E. coli has been studied. Anilazine delays the growth of E. coli by prolonging the lag period. It inhibits glucose oxidation by 60% and succinate oxidation by 100%. It
J Lewalter et al.
Toxicology letters, 107(1-3), 131-144 (1999-07-22)
The modern environmental awareness leads to the realisation that the human metabolism is stressed by a huge number of chemical substances. Generally, these background exposures, consisting predominantly from natural and partly from industrial as well as life style sources, are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service