Skip to Content
Merck
All Photos(1)

Documents

20801

Sigma-Aldrich

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate

meets analytical specification of Ph. Eur., BP, NF, 98-100.5% (calc with ref. to anhyd. subst.)

Synonym(s):

β,β,β-Trichloro-tert-butyl alcohol hemihydrate, ‘Acetone chloroform’, ‘Chlorobutanol’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CC(CH3)2OH · 0.5H2O
CAS Number:
Molecular Weight:
186.46
Beilstein:
878167
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

98-100.5% (calc with ref. to anhyd. subst.)

form

solid

quality

meets analytical specification of Ph. Eur., BP, NF

impurities

≤0.002% heavy metals (as Pb)
4.5-5.5% water (Karl Fischer)

ign. residue

≤0.1% (as SO4)

mp

76-80 °C

anion traces

chloride (Cl-): ≤100 mg/kg

application(s)

pharmaceutical (small molecule)

SMILES string

O.CC(C)(O)C(Cl)(Cl)Cl.CC(C)(O)C(Cl)(Cl)Cl

InChI

1S/2C4H7Cl3O.H2O/c2*1-3(2,8)4(5,6)7;/h2*8H,1-2H3;1H2

InChI key

WRWLCXJYIMRJIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate (chlorobutanol) is a preservative used as a standard for quantifying preservatives in pharmaceutical formulations by using capillary electrophoretic method. It has also been used to anesthetize leeches for preparing c-DNA library from regenerating retzius cells.

Biochem/physiol Actions

Chlorobutanol or 1,1,1-Trichloro-2-methyl-2-propanol is a opthalmic preservative and sedative hypnotic that possess antibacterial and antifungal properties. It also decreases the conduction velocity and induces conduction failure and automaticity within isolated ventricular muscle strips as well as impacts myocardial cells by acting on cell membrane and reduces isometric tension produced by heart.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Korneev et al.
Invertebrate neuroscience : IN, 3(2-3), 185-192 (1998-10-23)
We have constructed a subtractive cDNA library from regenerating Retzius cells of the leech, Hirudo medicinalis. It is highly enriched in sequences up-regulated during nerve regeneration. Sequence analysis of selected recombinants has identified both novel sequences and sequences homologous to
Elena S Novitskaya et al.
Clinics in dermatology, 29(3), 295-299 (2011-04-19)
Identifying contact allergens in ophthalmic medications can be a challenging and daunting experience. We summarize data on topical ophthalmic medications with the potential to cause periorbital contact dermatitis and allergic conjunctivitis, highlighting current dilemmas and controversies in this area. The
K Hermsmeyer et al.
The American journal of physiology, 230(2), 306-310 (1976-02-01)
The negative inotropic effect of a commonly used formulation of bradykinin (Sandoz BRS-640) was found to be due to chlorobutanol, a constituent of the preparation. Solutions containing up to 100 mug of crystalline bradykinin/ml had no effect on tension or
Małgorzata Jaworska et al.
Journal of separation science, 28(2), 137-143 (2005-03-10)
Preservatives are used to protect pharmaceutical formulations from microbial attack during the period of administration to the patient. Because of their biological activity, preservatives have to be identified and assayed according to the same rules as apply to active components.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service