Skip to Content
Merck
All Photos(1)

Key Documents

15678

Sigma-Aldrich

Boric anhydride

puriss. p.a., ≥98% (T)

Synonym(s):

Boron trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B2O3
CAS Number:
Molecular Weight:
69.62
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98% (T)
form:
solid
Pricing and availability is not currently available.

vapor density

>1 (vs air)

Quality Level

grade

puriss. p.a.

Assay

≥98% (T)

form

solid

loss

≤3% loss on ignition, 900 °C

mp

450 °C (lit.)

density

2.46 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤50 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤30 mg/kg
Mg: ≤20 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤30 mg/kg
Sr: ≤5 mg/kg
Tl: ≤10 mg/kg
Zn: ≤5 mg/kg

SMILES string

O=BOB=O

InChI

1S/B2O3/c3-1-5-2-4

InChI key

JKWMSGQKBLHBQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Boric anhydride (Boron trioxide) is a colorless glassy hygroscopic mass having rhombic crystals. Its dissolution in water is a highly exothermic reaction and orthoboric acid is formed. It can be prepared by heating boric acid. It is used as flux in metallurgical processes. It has been reported as an inhibitor of crystallization.[1]

Application

Boric anhydride can be used as a boron precursor to synthesize:
  • Boron carbon nitride nanosheet (BCN) by thermal condensation reaction with urea and melamine.
  • Boron-doped reduced graphene oxide (B-rGO) via a chemical vapor deposition method.
It can also be used as an additive along with pinacol to synthesize 4-alkoxy-5-hydroxy-2-pentenoate derivatives by Pd-catalyzed reaction of α, β-unsaturated γ, δ-epoxy esters with various alcohols.[2]

Other Notes

Reagent for the decomposition of silicates by fusion[3][4]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service