Skip to Content
Merck
All Photos(2)

Key Documents

MABT201

Sigma-Aldrich

Anti-Integrin alpha5 (Preservative Free) Antibody, clone SNAKA51

clone SNAKA51, from mouse, purified by using protein G

Synonym(s):

Integrin alpha-5, CD49 antigen-like family member E, Fibronectin receptor subunit alpha, Integrin alpha-F, VLA-5, CD49

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SNAKA51, monoclonal

purified by

using protein G

species reactivity

human

technique(s)

activity assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2α

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

General description

Integrins are a family of dimeric, transmembrane proteins that mediate cell-cell and extracellular matrix adhesion. Signals transduced by integrins play a role in many biological processes, including cell growth, differentiation, migration and apoptosis. The integrin family is composed of at least 15 alpha and 8 beta subunits that may form over twenty different alpha-beta non-covalently bound dimeric combinations on the cell surface. The alpha subunits all have some homology to each other, as do the beta subunits. Both of the subunits contribute to the binding of the ligand. Integrin alpha subunits contain seven weak sequence repeats in the N-terminal region, which may be important in ligand binding, and have been predicted to fold cooperatively into a single beta-propeller domain with seven beta-sheets. The α5/β1 integrin, also termed VLA5 or CD49e, is identical to the fibronectin receptor and mediates the adhesion of cells to fibronectin.

Immunogen

Whole cells corresponding to human Integrin alpha5.

Application

Activity Assay: A representative lot of this antibody converts alpha5 beta1 integrin into a ligand competant form and promotes fibronectin binding (Clark, K., et al. (2005). J Cell Sci. 118(Pt 2):291-300.).
Immunocytochemistry Analysis: A representative lot of this antibody detected Integin alpha 5 in Primary human foreskin fibroblasts (hFF) (Clark, K., et al. (2005). J Cell Sci. 118(Pt 2):291-300.; Valdembri, D., et al. (2009). PLoS Biol. 7(1):e25.).
Detect Integrin alpha5 (Preservative Free) using this Anti-Integrin alpha5 (Preservative Free) Antibody, clone SNAKA51 validated for use in Western Blotting, Activity Assay, ICC.
Research Category
Cell Structure

Quality

Evaluated by Flow Cytometry in HeLa cell treated with fibronectin.
Flow Cytometry Analysis: 1 µg of this antibody detected Integrin alpha5 in 0.5X10^6 HeLa cells.

Target description

116 kDa calculated

Physical form

Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Analysis Note

Control
HeLa cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bochuan Li et al.
The Journal of clinical investigation, 129(3), 1167-1179 (2019-01-11)
Local flow patterns determine the uneven distribution of atherosclerotic lesions. This research aims to elucidate the mechanism of regulation of nuclear translocation of Yes-associated protein (YAP) under oscillatory shear stress (OSS) in the atheroprone phenotype of endothelial cells (ECs). We
Erica Torchiaro et al.
Oncotarget, 7(1), 712-728 (2015-12-02)
The molecular mechanisms orchestrating peritoneal and hematogenous metastases of ovarian cancer cells are assumed to be distinct. We studied the p90RSK family of serine/threonine kinases that lie downstream the RAS-ERK/MAPK pathway and modulate a variety of cellular processes including cell
Nicole L Diggins et al.
Journal of cell science, 131(5) (2018-01-24)
Cell migration is a tightly coordinated process that requires the spatiotemporal regulation of many molecular components. Because adaptor proteins can serve as integrators of cellular events, they are being increasingly studied as regulators of cell migration. The adaptor protein containing
Mark S Filla et al.
Investigative ophthalmology & visual science, 60(12), 3897-3913 (2019-09-19)
Fibronectin fibrillogenesis is an integrin-mediated process that may contribute to the pathogenesis of primary open-angle glaucoma (POAG). Here, we examined the effects of αvβ3 integrins on fibrillogenesis in immortalized TM-1 cells and human trabecular meshwork (HTM) cells. TM-1 cells overexpressing
Gabrielle Larocque et al.
The Journal of cell biology, 220(10) (2021-07-22)
Membrane traffic is an important regulator of cell migration through the endocytosis and recycling of cell surface receptors such as integrin heterodimers. Intracellular nanovesicles (INVs) are transport vesicles that are involved in multiple membrane trafficking steps, including the recycling pathway.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service