Skip to Content
Merck
All Photos(2)

Key Documents

MABN2298M

Sigma-Aldrich

Anti-Prion Protein Antibody, clone POM2

clone POM2, from mouse

Synonym(s):

Major prion protein, PrP, PrP27-30, PrP33-35C, CD230

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

POM2, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Prnp(19122)

General description

Major prion protein (UniProt: P04925; also known as PrP, PrP27-30, PrP33-35C, CD230) is encoded by the Prnp (also known as Prn-p, Prp) gene (Gene ID: 19122) in murine species. PrP is highly expressed in the brain, lung, kidney and heart and low levels of expression are reported in the liver and spleen. It is found in high quantity in the brain of humans and animals infected with degenerative neurological diseases such as Kuru, Creutzfeldt-Jakob disease (CJD), Gerstmann-Straussler syndrome (GSS), scrapie, bovine spongiform encephalopathy (BSE), and transmissible mink encephalopathy (TME). In normal brain it may play a role in neuronal development and synaptic plasticity and may be required for neuronal myelin sheath maintenance and myelin homeostasis through acting as an agonist for ADGRG6 receptor. PrP can exist in either PrPc (c = cellular) or PrPsc (sc = Scrapie) form. PrPc is a glycoprotein normally found inserted in the plasma membrane by a glycosyl phosphatidylinositol (GPI) anchor and can be easily digested by proteases. It is composed mainly of alpha-helices. The PrPsc has more of a beta-sheet type structure and is highly resistant to the action of proteases. The prion proteins possess two N-linked glycosylation sites and may contain over 50 different sugars. The large size of the N-linked sugars enables them to shield two orthogonal faces of the protein almost completely and protect large regions of the protein surface from proteases. The PrPsc is shown to contain the same set of glycans as PrPc, but has a higher proportion of tri- and tetra-antennary sugars. Multiplication of prion proteins appears to be slower in vivo than in vitro. It is shown that following proteinase K treatment, PrPc is completely digested, whereas PrPsc is only shortened to a 27-30 kDa fragment. (Ref.: Meisl, G., et al. (2021). Nat. Struct. Mol. Biol. 28(4); 365-372).

Specificity

Clone POM2 detects major prion protein in Tga20 mice brain. It targets an epitope within the octapeptide repeat segment in the N-terminal region.

Immunogen

GST-tagged recombinant fragment corresponding to 209 amino acids from the internal region of murine major prion protein.

Application

Anti-Prion Protein, clone POM2, Cat. No. MABN2298, is a highly specific mouse monoclonal antibody that targets PrP and has been tested in ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: 0.25 ug/mL from a representative lot detected Prion Protein in mouse hippocampus tissue lysate.

Immunocytochemistry Analysis: Immunofluorescence Analysis: A representative lot detected Prion Protein in Immunocytochemistry applications (Kuffer, A., et. al. (2016). Nature. 536(7617):464-8).

ELISA Analysis: ELISA Analysis: A representative lot detected Prion Protein in ELISA applications (Polymenidou, M., et. al. (2008). PLoS One. 3(12):e3872).

Immunohistochemistry Analysis: A representative lot detected Prion Protein in Immunohistochemistry applications (Polymenidou, M., et. al. (2008). PLoS One. 3(12):e3872).

Immunoprecipitation Analysis: A representative lot detected Prion Protein in Immunoprecipitation applications (Kuffer, A., et. al. (2016). Nature. 536(7617):464-8; Polymenidou, M., et. al. (2008). PLoS One. 3(12):e3872).

Immunofluorescence Analysis: A representative lot detected Prion Protein in Immunofluorescence applications (Kuffer, A., et. al. (2016). Nature. 536(7617):464-8).

Western Blotting Analysis: A representative lot detected Prion Protein in Western Blotting applications (Kuffer, A., et. al. (2016). Nature. 536(7617):464-8; Polymenidou, M., et. al. (2008). PLoS One. 3(12):e3872).

Quality

Evaluated by Western Blotting in Brain homogenate from a Tga20 mouse overexpressing Prnp versus brain homogenate from a ZH3/ZH3 control mouse lacking Prnp.

Western Blotting Analysis: 0.05 µg/mL of this antibody this antibody detected PrP in Brain homogenate from a Tga20 mouse overexpressing Prnp versus brain homogenate from a ZH3/ZH3 control mouse lacking Prnp.


Target description

~35 kDa observed; 27.98 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service