Skip to Content
Merck
All Photos(2)

Key Documents

EZFITMIHE

Millipore

EZ-Fit® Manifold Filtration Head

For use with Microfil® filtration units, 1 unit or 3 units

Synonym(s):

Compatible with Microfil® filtration units, Microfil® Filtration head

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

316L stainless steel

Quality Level

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 1 unit (EZFITMIHE1)
pkg of 3 units (EZFITMIHE3)

manufacturer/tradename

EZ-Fit®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® (Filtration Manifold)

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

MICROFIL® filtration head is designed to improve ease of use, reduce maintenance and limit the risk of contamination. A continuous rim allows easy and safe membrane transfer with forceps, reducing the risk of touching the membrane filtration area. The porous filtration support can be removed simply by pressing down anywhere around its edge. These filtration heads offer full compatibility with the EZ-FIT® Filtration Units, stainless steel, and glass funnels.

Application

MICROFIL® filtration head is an accessory of the EZ-FIT® Manifold used during filtration of the test samples for general microbiological analysis, bioburden testing and water monitoring.

Packaging

EZFITMIHE1: 1 unit
EZFITMIHE3: 3 units
Comes with the porous membrane support EZFITFRIT3

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service