Skip to Content
Merck
All Photos(4)

Key Documents

ABN2244

Sigma-Aldrich

Anti-phospho-Parkin (Ser65)

purified antibody, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 51.64 kDa

species reactivity

human

species reactivity (predicted by homology)

bovine

packaging

antibody small pack of 100 μL

technique(s)

affinity binding assay: suitable
western blot: suitable

isotype

IgG

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer65)

Specificity

This rabbit polyclonal antibody specifically detects Parkin. It targets an epitope within 10 amino acids surrounding phosphoserine 65.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphoserine 65 from the N-terminal region of human parkin.

Application

Quality Control TestingEvaluated by Western Blotting in a lysate from a genetically modified proprietary cell line.Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected phosphorylated parkin (Ser 65) in a lysate from genetically modified cell line expressing phospho-Parkin (Ser 65).Tested ApplicationWestern Blotting Analysis: A 1:1,000 dilution from a representative lot detected phospho-Parkin (Ser65) in A431, C2C12, Hek293, and Hela cell lysates.Affinity Binding Assay: A representative lot of this antibody bound phospho-Parkin (Ser65) peptide with a KD of 1.0 x 10-12 in an affinity binding assay.Peptide Inhibition Analysis: A 1:1,000 dilution from a representative lot was used with Phosphorylated Parkin Ser65 lysate for peptide block analysis.Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-phospho-Parkin (Ser65), Cat. No. ABN2244, is a rabbit polyclonal antibody that detects E3 ubiquitin-protein ligase parkin and is tested for use in is tested for use in Affinity Binding Assay, Peptide Inhibition Assay, and Western Blotting.

Target description

E3 ubiquitin-protein ligase parkin (UniProt: O60260; also known as EC:2.3.2.31, Parkin, Parkin RBR E3 ubiquitin-protein ligase, Parkinson juvenile disease protein 2, Parkinson disease protein 2) is encoded by the PRKN (also known as PARK2) gene (Gene ID: 5071) in human. Parkin is mainly localized in the cytosol but relocates to dysfunctional mitochondria that have lost the mitochondrial membrane potential. Its mitochondrial recruitment is shown to be PINK1-dependent. Parkin functions within a multiprotein E3 ubiquitin ligase complex, catalyzing the covalent attachment of ubiquitin moieties onto various substrate proteins. It mediates monoubiquitination as well as polyubiquitination of substrates. It participates in the removal and/or detoxification of abnormally folded or damaged protein by mediating lysine 63-linked polyubiquitination of misfolded proteins. Parkin is also reported to mediate monoubiquitination of Bcl2 and acts as a positive regulator of autophagy and promotes the autophagic degradation of dysfunctional depolarized mitochondria by promoting the ubiquitination of TOMM20, RHOT1/MIRO1 and USP30. In its autoinhibited state the side chain of phenylalanine 463 inserts into a hydrophobic groove in RING-0 that occludes the ubiquitin acceptor site Cys-431. Its activation is a two-step process that involves serine 65 phosphorylation by PINK1 and binding to phosphorylated ubiquitin that leads to unlock repression of the catalytic Cys-431 by the RING-0 region via an allosteric mechanism and converting PRKN to its fully active form. Mutations in PRKN gene are known to cause Parkinson disease characterized by bradykinesia, resting tremor, muscular rigidity and postural instability.

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service