Skip to Content
Merck
All Photos(4)

Key Documents

ABE435

Sigma-Aldrich

Anti-trimethyl-Histone H3 (Lys36) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/I, Histone H3/j, Histone H3/k, Histone H3/l, H3K36me3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HIST1H3F(8968)

General description

Histones are nuclear proteins that form octameric structures which bind DNA to form units of chromatin called nucleosomes. The family of histones—H2A, H2B, H3, and H4—are key players in gene regulation. They undergo a number of post-translational modifications (PTM) in response to various stimuli including phosphorylation on serine and threonine residues and methylation on lysine residues. PTMs produce configural changes in histone proteins that may induce nucleosome remodeling and expose or hide DNA sequences from transcriptional complexes. Methylation occurs on histone H3 at lysine 4, 9, 27, 36, and 79 residues. Previous studies have reported that methylation of H3 lysine 9 provides a binding site for heterochromatin adaptor proteins (HP1) which are involved in the formation of heterochromatin and gene repression. Phosphorylation of histones may also produce similar repressive effects depending on the cellular context. Often, it is the pattern of different modifications on several histone residues that alter chromatin, rather than modification of single residues.

Specificity

Broad species reactivity expected due to similar sequence homology.
This antibody recognizes Histone H3 trimethylated at Lys36.

Immunogen

Epitope: Trimethylated Lys36
KLH-conjugated linear peptide corresponding to human Histone H3 trimethylated at Lys36.

Application

Anti-trimethyl-Histone H3 (Lys36) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated at lysine 36. Also known as Anti-H3K36me3, this purified Ab is specificity verified by dot blot (DB) and has been validated in WB, ICC, DB, ChIP, ChIP-seq.
Dot Blot Specificity Analysis: Unmodified and various modified Histone peptides (see Table) were probed with Anti-trimethyl-Histone H3 (Lys36) (1:2,000 dilution).

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Histone H3 in A431 cells.

ChIP-Sequencing: Chromatin immunoprecipitation was performed using the Magna ChIP HiSens kit (cat#17-10460, 2 µg of anti-trimethyl-Histone H3 (Lys36) antibody(ABE435), 20 µL Protein A/G beads , and 1e6 crosslinked HeLa cell chromatin followed by DNA purification using magnetic beads. Libraries were prepared from Input and ChIP DNA samples using standard protocols with Illumina barcoded adapters, and analyzed on Illumina HiSeq instrument. An excess of eleven million reads from FastQ files were mapped using Bowtie (http://bowtie-bio.sourceforge.net/manual.shtml) following TagDust (http://genome.gsc.riken.jp/osc/english/dataresource/) tag removal. Peaks were identified using MACS (http://luelab.dfci.harvard.edu/MACS/), with peaks and reads visualized as a custom track in UCSC Genome Browser (http://genome.ucsc.edu) from BigWig and BED files.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Western Blot in HeLa cell extract and recombinant Histone H3 protein.

Western Blot Analysis: A 1:500 dilution of this antibody detected Histone H3 on 10 µg of HeLa cell extract and recombinant Histone H3 protein.

Target description

~17 kDa observed. An uncharacterized band appears at ~52 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell extract and recombinant Histone H3 protein

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandip De et al.
Science advances, 5(1), eaau9739-eaau9739 (2019-01-22)
Polycomb group proteins (PcGs) drive target gene repression and form large chromatin domains. In Drosophila, DNA elements known as Polycomb group response elements (PREs) recruit PcGs to the DNA. We have shown that, within the invected-engrailed (inv-en) Polycomb domain, strong
Sandip De et al.
Genetics, 216(3), 689-700 (2020-09-20)
Polycomb group (PcG) proteins are an important group of transcriptional repressors that act by modifying chromatin. PcG target genes are covered by the repressive chromatin mark H3K27me3. Polycomb repressive complex 2 (PRC2) is a multiprotein complex that is responsible for
Huijuan Song et al.
Cancer science, 111(10), 3665-3678 (2020-08-04)
Androgen receptor (AR) signaling is considered to be crucial for the pathogenesis of hepatocellular carcinoma (HCC) with obvious sexual dimorphism. Pre-mRNA processing factor 6 (PRPF6) was identified as a coactivator of AR. However, the molecular mechanism underlying the modulation function
Philipp Tropberger et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(42), E5715-E5724 (2015-10-07)
Chronic hepatitis B virus (HBV) infection affects 240 million people worldwide and is a major risk factor for liver failure and hepatocellular carcinoma. Current antiviral therapy inhibits cytoplasmic HBV genomic replication, but is not curative because it does not directly
Wei Liu et al.
International journal of biological sciences, 17(1), 188-203 (2021-01-05)
Androgen receptor (AR) and its variants play vital roles in development and progression of prostate cancer. To clarify the mechanisms involved in the enhancement of their actions would be crucial for understanding the process in prostate cancer and castration-resistant prostate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service