8.52324
Fmoc-bishomopropargylglycine
Novabiochem®
Synonym(s):
Fmoc-bishomopropargylglycine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C22H21NO4
Molecular Weight:
363.41
UNSPSC Code:
12352209
NACRES:
NA.22
Recommended Products
Quality Level
product line
Novabiochem®
form
solid
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
alkyne
storage temp.
15-25°C
Related Categories
General description
Building block for the introduction by Fmoc SPPS of an amino acid bearing an alkyne side chain. Such derivatives are useful tools for the synthesis of cyclic peptides by an alkyne-alkyne Glaser coupling, involving Cu(OAc)2/pyridine and heating to 60 °C using microwaves.
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Guide to Selection of Orthogonally -Protected Building Blocks for Fmoc SPPS
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Guide to Selection of Orthogonally -Protected Building Blocks for Fmoc SPPS
Analysis Note
Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service