8.41036
Decane-1-sulfonic acid sodium salt
for synthesis
Synonym(s):
Decane-1-sulfonic acid sodium salt, Sodium 1-decanesulphonate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H21NaO3S
CAS Number:
Molecular Weight:
244.33
MDL number:
UNSPSC Code:
12352107
EC Index Number:
236-525-9
NACRES:
NA.22
Quality Level
form
powder
mp
>300 °C
storage temp.
2-30°C
SMILES string
[Na+].[S](=O)(=O)([O-])CCCCCCCCCC
InChI
1S/C10H22O3S.Na/c1-2-3-4-5-6-7-8-9-10-14(11,12)13;/h2-10H2,1H3,(H,11,12,13);/q;+1/p-1
InChI key
AIMUHNZKNFEZSN-UHFFFAOYSA-M
Analysis Note
Assay (acidimetric after ion exchange): ≥ 98.0 %
Identity (IR): passes test
Identity (IR): passes test
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service