Skip to Content
Merck
All Photos(1)

Key Documents

8.21100

Sigma-Aldrich

Pyrrolidine-1-dithiocarboxylic acid ammonium salt

for synthesis

Synonym(s):

Pyrrolidine-1-dithiocarboxylic acid ammonium salt, Ammonium-N,N-tetramethylenedithiocarbamate, N,N-Tetramethylenedithiocarbaminic acid ammoniumsalt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2S2
CAS Number:
Molecular Weight:
164.29
MDL number:
UNSPSC Code:
12352107
EC Index Number:
225-834-4
NACRES:
NA.22
grade:
for synthesis
form:
solid
solubility:
190 g/L
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

form

solid

pH

6-7 (20 °C, 50 g/L in H2O)

mp

146-151 °C

solubility

190 g/L

bulk density

400 kg/m3

storage temp.

2-8°C

SMILES string

[S-]C(=S)N1CCCC1.[N+H4]

InChI

1S/C5H9NS2.H3N/c7-5(8)6-3-1-2-4-6;/h1-4H2,(H,7,8);1H3

InChI key

MDDIUTVUBYEEEM-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service