Skip to Content
Merck
All Photos(1)

Key Documents

8.18061

Sigma-Aldrich

Diphenyl diselenide

for synthesis

Synonym(s):

Diphenyl diselenide, Phenyl diselenide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10Se2
CAS Number:
Molecular Weight:
312.13
MDL number:
UNSPSC Code:
12352103
EC Index Number:
216-780-2
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0% (GC)

form

powder

mp

58-61 °C

storage temp.

2-30°C

SMILES string

[Se]([Se]c2ccccc2)c1ccccc1

InChI

1S/C12H10Se2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10H

InChI key

YWWZCHLUQSHMCL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 58 °C
Melting range (upper value): ≤ 61 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service