Skip to Content
Merck
All Photos(2)

Key Documents

8.06022

Sigma-Aldrich

Methanesulfonic acid

for synthesis

Synonym(s):

Methanesulfonic acid, MSA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH4O3S
CAS Number:
Molecular Weight:
96.11
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-898-6
NACRES:
NA.22

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

<1 hPa ( 20 °C)

Assay

≥99.0% (acidimetric)

form

liquid

potency

649 mg/kg LD50, oral (Rat)
200-2000 mg/kg LD50, skin (Rabbit)

bp

167 °C/13 hPa

mp

20 °C

transition temp

flash point 189 °C

solubility

1000 g/L

density

1.48 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[S](=O)(=O)([O-])C.[H+]

InChI

1S/CH4O3S/c1-5(2,3)4/h1H3,(H,2,3,4)

InChI key

AFVFQIVMOAPDHO-UHFFFAOYSA-N

Application

  • Methanesulfonic Acid in Hydrometallurgy: This paper reviews the properties of methanesulfonic acid and its applications in hydrometallurgy, highlighting its benefits over other acids. (Binnemans and Jones, 2023).
  • The Role of MSA in Aerosol Formation and Growth: This study investigates how MSA can influence aerosol formation and the associated radiative forcings, emphasizing its environmental impacts. (Hodshire et al., 2019).
  • Organic Acids in New Particle Formation: Research on the role of organic acids in enhancing MSA-driven new particle formation, providing insights into atmospheric chemistry. (Zhang et al., 2022).
  • Methanesulfonic Acid as a Sustainable Acidic Solvent: This article discusses the use of MSA for recovering metals from zinc industry residue, showcasing its applications in sustainable metallurgy. (Palden et al., 2019).

Analysis Note

Appearance of substance (visual): colourless to brownish
Assay (acidimetric): ≥ 99.0 %
Density (d 20 °C/ 4 °C): 1.478 - 1.482
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

372.2 °F - closed cup

Flash Point(C)

189 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service