Skip to Content
Merck
All Photos(2)

Key Documents

8.04393

Sigma-Aldrich

1-Hexanol

for synthesis

Synonym(s):

n-Hexyl alcohol, Hexyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5OH
CAS Number:
Molecular Weight:
102.17
Beilstein:
969167
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-852-3
NACRES:
NA.22
Assay:
≥98% (GC)
bp:
156-157 °C (lit.)
vapor pressure:
1 mmHg ( 25.6 °C)
Pricing and availability is not currently available.

vapor density

4.5 (vs air)

Quality Level

vapor pressure

1 mmHg ( 25.6 °C)

Assay

≥98% (GC)

form

liquid

autoignition temp.

559 °F

potency

>2000 mg/kg LD50, skin (Rabbit)

refractive index

n20/D 1.418 (lit.)

kinematic viscosity

3.64 cSt(40 °C)

bp

156-157 °C (lit.)

mp

−52 °C (lit.)

transition temp

flash point 60 °C

density

0.814 g/mL at 25 °C (lit.)

storage temp.

no temp limit

SMILES string

CCCCCCO

InChI

1S/C6H14O/c1-2-3-4-5-6-7/h7H,2-6H2,1H3

InChI key

ZSIAUFGUXNUGDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Fast Synthesis and Photoluminescent Property Regulation of Carbon Quantum Dots: Describes the use of 1-hexanol as a precursor in the synthesis of carbon quantum dots, impacting their photoluminescent properties (Y Li et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 98 %
Density (d 20 °C/ 4 °C): 0.818 - 0.819
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service