8.02100
Calcium hydride
for synthesis
Synonym(s):
Calcium hydride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
H2Ca
CAS Number:
Molecular Weight:
42.09
MDL number:
UNSPSC Code:
12352306
EC Index Number:
232-189-2
NACRES:
NA.22
Quality Level
form
solid
autoignition temp.
>300 °C
reaction suitability
reagent type: reductant
mp
816 °C
density
1.9 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
[Ca]
InChI
1S/Ca
InChI key
OYPRJOBELJOOCE-UHFFFAOYSA-N
Related Categories
Application
Calcium hydride (CaH2) can be used as a reducing agent to synthesize metallic elements (e.g., Ti, Zr, V, Nb, U, and Th) from metal oxides via the calcium hydride reduction process at higher temperature (600–1000°C). Metallic calcium produced by thermal decomposition is the actual reducing agent in this process. CaH2 is also used to reduce various metal hydrides to metals.
Analysis Note
Assay (by gasvolumetry): ≥ 95.0 %
Particle size: 0 -20 mm
Particle size: 0 -20 mm
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Water-react 1
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Hydrides
Rittmeyer P and Wietelmann U
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service