Skip to Content
Merck
All Photos(1)

Key Documents

8.00832

Sigma-Aldrich

Butyl acrylate

(stabilised) for synthesis

Synonym(s):

Butyl acrylate, Acrylic acid butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCOO(CH2)3CH3
CAS Number:
Molecular Weight:
128.17
MDL number:
UNSPSC Code:
12352108
EC Index Number:
205-480-7
NACRES:
NA.22
grade:
for synthesis
bp:
147-148 °C/1013 hPa
vapor pressure:
29 hPa ( 50 °C)
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

vapor pressure

29 hPa ( 50 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

275 °C

potency

3140 mg/kg LD50, oral (Rat)

expl. lim.

1.1-7.8 % (v/v)

dilution

(for synthesis)

bp

147-148 °C/1013 hPa

mp

-64.6 °C

transition temp

flash point 38 °C

solubility

1.7 g/L

density

0.90 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCCC)C(=O)C=C

InChI

1S/C7H12O2/c1-3-5-6-9-7(8)4-2/h4H,2-3,5-6H2,1H3

InChI key

CQEYYJKEWSMYFG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.898 - 0.899
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service