Skip to Content
Merck
All Photos(1)

Key Documents

489479

Sigma-Aldrich

LSD1 Inhibitor IV, RN-1, 2HCl

LSD1 Inhibitor IV, RN-1, HCl, is a cell-permeable potent, irreversible inhibitor of lysine specific demethylase 1 (LSD1; IC₅₀ = 70 nM in a HRP-coupled assay using H3K4Me2 peptide substrate).

Synonym(s):

LSD1 Inhibitor IV, RN-1, 2HCl, LSD Inhibitor IV, Histone Lysine Demethylase Inhibitor VI, MOA Inhibitor III, 2-(1R,2S)-2-(4-(Benzyloxy)phenyl)cyclopropylamino)-1-(4-methylpiperazin-1-yl)ethanone, HCl, BHC110 Inhibitor IV, KDM1 Inhibitor IV, LSD Inhibitor IV, KDM1 Inhibitor IV, Histone Lysine Demethylase Inhibitor VI, MOA Inhibitor III, 2-(1R,2S)-2-(4-(Benzyloxy)phenyl)cyclopropylamino)-1-(4-methylpiperazin-1-yl)ethanone, HCl, BHC110 Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H29N3O2 · 2HCl
Molecular Weight:
452.42
UNSPSC Code:
12352208
NACRES:
NA.28

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 2.5 mg/mL
water: 2.5 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable tranylcypromine (parnate; Cat. No. 616431) analog that acts as a potent, irreversible inhibitor of lysine specific demethylase 1 (LSD1; IC50 = 70 nM in a HRP-coupled assay using H3K4Me2 peptide substrate). Forms a covalent adduct with flavin adenine dinucleotide (FAD). Shown to cross the blood brain barrier. Exhibits moderate selectivity over amine oxidases MAO-A and MAO-B (IC50 = 0.51 and 2.785 µM, respectively). Displays desirable pharmacokinetic properties (brain/plasma exposure ratio of 88.9), and impairs long-term memory without affecting short-term memory in mice (10 mg/kg, i.p.).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Neelamegam, R., et al. 2011. ACS Chem. Neurosci.3, 120.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoying Zhou et al.
Frontiers in cell and developmental biology, 10, 861916-861916 (2022-08-09)
Commensal microbes cross talk with their colonized mucosa. We show that microbes and their cell wall components induce an inflammatory response in cultured human mucosal cells derived from the nonmalignant nasopharyngeal epithelium (NNE) cells in vitro. NNE cells show significant
Brennan D Johnson et al.
ACS bio & med chem Au, 2(2), 161-170 (2022-07-28)
The flavin adenine dinucleotide containing Endoplasmic Reticulum Oxidoreductase-1 α (ERO1α) catalyzes the formation of de novo disulfide bond formation of secretory and transmembrane proteins and contributes towards proper protein folding. Recently, increased ERO1α expression has been shown to contribute to
Ramesh Neelamegam et al.
ACS chemical neuroscience, 3(2), 120-128 (2012-07-04)
Modulation of histone modifications in the brain may represent a new mechanism for brain disorder therapy. Post-translational modifications of histones regulate gene expression, affecting major cellular processes such as proliferation, differentiation, and function. An important enzyme involved in one of
Amanda Holley et al.
Physiology & behavior, 196, 78-83 (2018-08-24)
Although female rats are typically described as having a promiscuous mating strategy, if sexually naïve females have their formative sexually rewarding experiences paired with the same male, they will recognize that male and display mate-guarding behavior towards him in the
Cellular population dynamics shape the route to human pluripotency.
Panariello, et al.
Nature Communications, 14, 2829-2829 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service