444180
MAP Kinase Inhibitor Set I
The MAP Kinase Inhibitor Set I controls the biological activity of MAP Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
solubility
DMSO: soluble
shipped in
ambient
storage temp.
−20°C
General description
Contains 5 mg of the MEK inhibitor PD 98059 (Cat. No. 513000), 1 mg each of the MAP kinase inhibitors SB 202190 (Cat. No. 559388) and SB 203580 (Cat. No. 559389), and 1 mg of the negative control, SB 202474 (Cat. No. 559387). Supplied with a data sheet.
MAP (mitogen-activated protein) kinases are ubiquitous serine/threonine kinases that are activated by a wide variety of extracellular signals. The MAP kinase cascade plays a crucial role in signal transduction from the membrane to the nucleus and appears to be highly conserved throughout evolution. In mammalian cells, this cascade is activated by both tyrosine kinase receptors and G-protein coupled receptors. MAP kinase activation, an essential step in triggering cell cycle progression, leads to changes in properties of cytoplasmic, membrane-associated and nuclear proteins. A unique feature of MAP kinases is that their full activation requires phosphorylation of both a threonine residue and a tyrosine residue which are separated by a single amino acid. This phosphorylation is brought about by the action of MAP kinase kinase (MEK), a dual specificity kinase. Once activated, MAP kinase relays signals downstream by phosphorylating various other protein kinases and regulatory proteins. Following activation, MAP kinases translocate from the cytosol into the nucleus and phosphorylate a number of transcription factors leading to changes in gene expression. Supplied with a data sheet.
Biochem/physiol Actions
Primary Target
p38 kinase
p38 kinase
Target IC50: 350 nM, 600 nM
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, aliquot and store at -20°C.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service