Skip to Content
Merck
All Photos(2)

Key Documents

36-008-C

Sigma-Aldrich

Anti-α-Synuclein (SNCA) Antibody

mouse monoclonal, Syn211

Synonym(s):

Alpha-synuclein, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, NACP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-alpha-Synuclein (aa 121-125) Antibody, clone Syn211 (Ascites Free), clone Syn211, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Syn211, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SNCA(6622)

General description

Alpha-synuclein is encoded by the SNCA gene (also known as NACP, PARK1) in human. There are two other known synuclein family members, beta- and gamma-synuclein. Interest in synuclein research started with the discovery of alpha-synuclein mutation in several families with autosomal dominant Parkinson′s disease (PD). Alpha-synuclein is abundantly expressed in the brain and is found in a classic amyloid fibril form within the intra-neuronal Lewy body deposits of PD. The physiological function of synucleins is not well understood, but appears to involve membrane interactions, and in particular reversible binding to synaptic vesicle membranes. Contradictory evidences regarding inhibitory action of alpha-synuclein against phospholipase D (PLD) have been reported (PMID 11821392 & 19146388). Clone Syn211, Cat. No. 36-008-C detects epitope in the region from amino acid 121 to 125 and has been used together with clone LB509, Cat. No. MABN824, to characterize the digestion pattern of different forms of in vitro prepared alpha-synclulein aggregates (Guo, J.L., et al. 2013; PMID 23827677).

Immunogen

Conjugated recombinant protein corresponding to human alpha-Synuclein.
Epitope: alpha-syn 121-125

Application

Anti-alpha-Synuclein (aa 121-125) Antibody, clone Syn211 (Ascites Free) is an antibody against alpha-Synuclein for use in Immunohistochemistry and Western Blotting.
Immunohistochemistry Analysis: A 1:50-1,000 dilution from a representative lot detected alpha-Synuclein in human cerebral cortex and human kidney tissue.
Western Blotting Analysis: A representative lot detected alpha-Synuclein in Synucleinopathy brains (Guo, J.L., et al. (2013). Cell. 154:103-117).

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected alpha-Synuclein in 10 µg of human brain tissue lysate.

Target description

~15 kDa observed

Physical form

Format: Purified

Storage and Stability

Stable for 1 year at 2-8°Cfrom date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Distinct ?-synuclein strains differentially promote tau inclusions in neurons.
Guo, JL; Covell, DJ; Daniels, JP; Iba, M; Stieber, A; Zhang, B; Riddle, DM; Kwong, LK; Xu et al.
Cell null
Kyoung-In Cho et al.
Cellular and molecular life sciences : CMLS, 76(17), 3407-3432 (2019-04-05)
Nucleocytoplasmic transport is dysregulated in sporadic and familial amyotrophic lateral sclerosis (ALS) and retinal ganglion neurons (RGNs) are purportedly involved in ALS. The Ran-binding protein 2 (Ranbp2) controls rate-limiting steps of nucleocytoplasmic transport. Mice with Ranbp2 loss in Thy1+-motoneurons develop

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service