Skip to Content
Merck
All Photos(1)

Key Documents

345804

Sigma-Aldrich

β1-3,6-Galactosidase, Recombinant, E. coli

Synonym(s):

β1-3,6-Galactosidase, Recombinant, E. coli, β1-3,6-D-Galactoside Galactohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

form

liquid

specific activity

≥30 units/mL
≥75 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

Proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

General description

Recombinant, β1-3,6-Galactosidase expressed in E. coli. Catalyzes the hydrolysis of non-reducing terminal β1,3- and β1,6-linked galactose from complex carbohydrates and glycoproteins. Not known to cleave substituted or branched galactose. Inhibited by fucose attached to penultimate sugar.
Recombinant, β1-3,6-Galactosidase expressed in E. coli. Catalyzes the hydrolysis of non-reducing terminal β1,3-and β1,6-linked galactose from complex carbohydrates and glycoproteins. Not known to cleave substituted or branched galactose. Inhibited by fucose attached to penultimate sugar.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will catalyze the release of 1.0 µmol p-nitrophenol from p-nitrophenyl-β-D-galactopyranoside per min at 37°C, pH 5.0.

Physical form

In 25 mM NaCl, 20 mM Tris-HCl, pH 7.5.

Other Notes

Prime, S., et al. 1996. J. Chromatogr. A. 720, 263.
Dwek, R.A., et al. 1993. Annu. Rev. Biochem. 62, 65.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabella Faraoni et al.
Cancers, 11(9) (2019-09-19)
Myelodysplastic syndromes (MDS) are highly heterogeneous myeloid diseases, characterized by frequent genetic/chromosomal aberrations. Olaparib is a potent, orally bioavailable poly(ADP-ribose) polymerase 1 (PARP1) inhibitor with acceptable toxicity profile, designed as targeted therapy for DNA repair defective tumors. Here, we investigated
Huang Zhu et al.
Cell stem cell, 27(2), 224-237 (2020-06-13)
Cytokine-inducible SH2-containing protein (CIS; encoded by the gene CISH) is a key negative regulator of interleukin-15 (IL-15) signaling in natural killer (NK) cells. Here, we develop human CISH-knockout (CISH-/-) NK cells using an induced pluripotent stem cell-derived NK cell (iPSC-NK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service