Skip to Content
Merck
All Photos(1)

Key Documents

23-036-M

Sigma-Aldrich

UbcH6 Protein, active, 10 µg

Active, full length, recombinant human UbcH6. N-terminal HA, 6His-tagged. For use in Enzyme Assays. Functions as an E2 enzyme in ubiquitination assays.

Synonym(s):

Ubiquitin-conjugating enzyme E2 E1, Ubiquitin-protein ligase E1, UBE2E1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

human

Quality Level

100
300

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

NCBI accession no.

UniProt accession no.

General description

Ubiquitin-conjugating (E2) enzymes are characterized by the presence of a highly conserved ubiquitin-conjugating domain which accommodates ATP-activated ubiquitin (Ub) via a covalently linked thioester onto its active-site residue. E2 enzymes act via selective protein-protein interactions with the ubiquitin-activating E1 enzyme and ubiquitin ligase E3 enzymes and are able to differentiate effects on downstream substrates, either with a single Ub molecule or a Ub chain. While E3s are involved in substrate selection, E2s are the main determinants for selection of the lysine to construct Ub chains, which thereby directly control the cellular fate of the substrate.
UbcH6 has been shown to interact with a number of diverse E3 ligases, including RNF20/40, Trim32 and WWP2.

Application

Active, full length, recombinant human UbcH6. N-terminal HA, 6His-tagged. For use in Enzyme Assays. Functions as an E2 enzyme in ubiquitination assays.
Ubiquitination Cascade Comoponent: E2

Physical form

Purified using immobilized metal affinity chromatography.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service