Skip to Content
Merck
All Photos(1)

Key Documents

219007

Sigma-Aldrich

Caspase Inhibitor VI

Z-VAD-FMK, CAS 161401-82-7, is an irreversible pan caspase inhibitor. Does not require pretreatment with esterase for in vitro studies.

Synonym(s):

Caspase Inhibitor VI, Z-VAD-FMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28FN3O7
CAS Number:
Molecular Weight:
453.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

off-white

solubility

DMSO: 9 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

FCC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)OCc1ccccc1)C(C)C)C)CC(=O)O

InChI

1S/C21H28FN3O7/c1-12(2)18(25-21(31)32-11-14-7-5-4-6-8-14)20(30)23-13(3)19(29)24-15(9-17(27)28)16(26)10-22/h4-8,12-13,15,18H,9-11H2,1-3H3,(H,23,30)(H,24,29)(H,25,31)(H,27,28)/t13-,15-,18-/m0/s1

InChI key

SUUHZYLYARUNIA-YEWWUXTCSA-N

General description

An irreversible general caspase inhibitor. Useful for studies involving recombinant, isolated, and purified caspase enzymes. Unlike Caspase Inhibitor I (Cat. No. 627610), this inhibitor does not require pretreatment with esterase for in vitro studies. A 10 mM (1 mg/221 µl) solution of Caspase Inhibitor VI (Cat. No. 219011) in DMSO is also available.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiara Brignola et al.
Antioxidants (Basel, Switzerland), 13(7) (2024-07-27)
Despite advancements in therapeutic strategies, the development of drug resistance and metastasis remains a serious concern for the efficacy of chemotherapy against colorectal cancer (CRC). We have previously demonstrated that low expression of ribosomal protein uL3 positively correlates with chemoresistance
Xiaoqing Qian et al.
Journal of cellular physiology, 236(7), 5235-5252 (2020-12-29)
Intratympanic injection of gentamicin has proven to be an effective therapy for intractable vestibular dysfunction. However, most studies to date have focused on the cochlea, so little is known about the distribution and uptake of gentamicin by the counterpart of
Wayne A Cabral et al.
Aging cell, 20(9), e13457-e13457 (2021-08-29)
Hutchinson-Gilford progeria syndrome (HGPS) is a rare accelerated aging disorder most notably characterized by cardiovascular disease and premature death from myocardial infarction or stroke. The majority of cases are caused by a de novo single nucleotide mutation in the LMNA
Amphipathic barbiturates as marine product mimics with cytolytic and immunogenic effects on head and neck squamous cell carcinoma cell lines.
von Hofsten, et al.
Frontiers in Pharmacology, 14, 1141669-1141669 (2023)
Takashi Kojima et al.
Disease models & mechanisms, 14(11) (2021-10-09)
KCNQ4 encodes the homotetrameric voltage-dependent potassium ion channel Kv7.4, and is the causative gene for autosomal dominant nonsyndromic sensorineural hearing loss, DFNA2. Dominant-negative inhibition accounts for the observed dominant inheritance of many DFNA2-associated KCNQ4 variants. In addition, haploinsufficiency has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service