If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
1.88010
Titrant 5
1 ml ≙ ca. 5 mg H₂O, titrant for Karl Fischer titration with two component reagents, Aquastar®
About This Item
Recommended Products
Product Name
Titrant 5, titrant for volumetric Karl Fischer titration with two component reagents 1 ml ≙ ca. 5 mg H₂O Aquastar®
Quality Level
product line
Aquastar®
form
liquid
reaction suitability
reaction type: volumetric
concentration
5 mg/mL
technique(s)
Karl-Fischer titration: suitable
titration: suitable
transition temp
flash point <21 °C
density
0.85 g/cm3 at 20 °C
format
two component solution
storage temp.
15-25°C
Related Categories
Packaging
Analysis Note
Other Notes
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT RE 2 Oral - STOT SE 1
Target Organs
Thyroid
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F - (Methanol)
Flash Point(C)
9.7 °C - (Methanol)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Summary application report for analysis of moisture in D(+)-glucose-monohydrate
Summary application report for analysis of moisture in Ammonia, aqueous
Summary application report for analysis of moisture in D(+)-glucose-monohydrate
Summary application report for analysis of moisture in D(+)-glucose-monohydrate
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
-
Is the titrant 5 1.88010 for Karl Fischer contains both the titrant and solvent needed for Karl Fischer as it shows in picture.
1 answer-
Product 1880101045 only has the Titrant 5.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service