Skip to Content
Merck
All Photos(1)

Key Documents

1.74011

Supelco

Rectangular cell 100 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
SE.11

material

optical glass vial

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Prove 600

detection method

photometric

storage temp.

no temp limit

General description

The sensitivity of a measurement is directly linked to the path length of the cuvette that is used in the photometeric measurement. Using this 100 mm rectangular cuvette is possible in our Spectroquant Prove 600. It allows to measure very low concentrations of e.g. bromide or phosphate in water, drinking water, ground water, and surface water. Please consider our application notes for more information on this.

The rectangular cuvette comes with an instruction sheet on how to use the cuvette.
for Prove 600 Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spectroquant® photometric test accurately quantifies low concentrations of PO4-P in natural waters.

Spectroquant® photometric test accurately quantifies low concentrations of PO4-P in natural waters.

Spectroquant® photometric test accurately quantifies low concentrations of PO4-P in natural waters.

Spectroquant® photometric test accurately quantifies low concentrations of PO4-P in natural waters.

See All

Protocols

Photometric determination of phosphate ions in water with ammonium molybdate using Spectroquant® test kits and a UV/VIS spectrophotometer.

Photometric determination of phosphate ions in water with ammonium molybdate using Spectroquant® test kits and a UV/VIS spectrophotometer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service