Skip to Content
Merck
All Photos(1)

Key Documents

12-302

Sigma-Aldrich

EGF-Stimulated A431 Cell Lysate

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105330
eCl@ss:
32011807
NACRES:
NA.41

product name

EGF-Stimulated A431 Cell Lysate,

biological source

human

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

shipped in

dry ice

Cell Line Description

A431

Quality

Routinely evaluated as a positive control for many of Upstates antibodies.

Physical form

modified RIPA buffer (50mM Tris-HCl, pH 7.4, 1% NP40, 0.25% sodium deoxycholate, 150mM NaCl, 1mM EDTA, 1mM PMSF, 1mg/ml aprotinin, 1mg/ml leupeptin, 1mg/ml pepstatin, 1mM Na3VO4, 1mM NaF) and diluted with non-reducing sample buffer (31mM Tris-HCl, pH 6.8, 5% glycerol, 1% SDS, 0.002% bromophenol blue).

Storage and Stability

6 months at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Svetoslava S Kerpedjieva et al.
Stem cells and development, 21(13), 2541-2551 (2012-02-10)
Cell therapy with adult bone marrow multipotential stromal cells/mesenchymal stem cells (MSCs) presents a promising approach to promote wound healing and tissue regeneration. The strong paracrine capability of various growth factors and cytokines is a key mechanism of MSC-mediated wound
Narendra Padhan et al.
BMC cancer, 16(1), 683-683 (2016-08-27)
The progression of colorectal cancer (CRC) involves recurrent amplifications/mutations in the epidermal growth factor receptor (EGFR) and downstream signal transducers of the Ras pathway, KRAS and BRAF. Whether genetic events predicted to result in increased and constitutive signaling indeed lead
Srinivasan Shanmugarajan et al.
Endocrinology, 151(9), 4389-4399 (2010-07-09)
Osteoclast inhibitory peptide-1 (OIP) is an autocrine/paracrine inhibitor of osteoclast differentiation, and mice that overexpress OIP-1 in osteoclast lineage cells develop an osteopetrosis bone phenotype. In this study, we show that OIP-1 binding to the Fc gamma receptor IIB (Fc
Endrit Elbasani et al.
Journal of virology, 94(13) (2020-04-17)
The anaphase-promoting complex, or cyclosome (APC/C), is a large E3 ubiquitin ligase composed of 14 subunits. The activity of APC/C oscillates during the cell cycle to ensure a timely transition through each phase by promoting the degradation of important cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service