Skip to Content
Merck
All Photos(4)

Key Documents

1.17421

Millipore

Safe Lid for MAS-100 NT®

new

for use with MAS-100 NT®, sterile; irradiated (Alternative to the standard autoclavable perforated lid), ready-to-use, box of 30 units

Synonym(s):

Ready-to use Perforated Lid, Sterile Perforated Lid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103403
NACRES:
NA.74

material

acrylonitrile-butadiene-styrene (ABS) lid

Quality Level

sterility

sterile; irradiated (Alternative to the standard autoclavable perforated lid)

feature

disposable
ready-to-use

packaging

box of 30 units

manufacturer/tradename

MAS-100 NT®

application(s)

pharmaceutical

compatibility

for use with MAS-100 NT®

General description

Safe Lid for MAS-100 NT® is a disposable alternative to the autoclavable standard perforated lid made from aluminum (ref. 1.09195). It is made of 100% recyclable acrylonitrile butadiene styrene (ABS) and comes in 30 individually double-packed and sterilized pieces per package.

Safe Lid has the same sieve configuration as the standard perforated lid with 300 holes of 0.6 mm diameter. This ensures an identical impact velocity of 19.6 m/s and a nominal d50 of 1.1 μm. The simplified validation of the recovery rates according to EN 17141 resulted in comparable values for both perforated lids.

Application

The application of the disposable lid on the MAS-100 NT® air sampler is identical to that of the standard perforated lid. You may follow the user instructions of MAS-100 NT®. The only difference is the lack of a twist fastener, which simplifies handling even more.

Features and Benefits

  • Collection efficiency of ISO 14698 validated MAS-100 NT® is comparable for standard perforated lids and Safe Lid (verified by simplified validation acc. EN 17141).
  • Reproducible results thanks to 100% visual, camera-based control of the holes.
  • Flow rate with MAS-100 NT® is 100 SLPM ± 5% .
  • By using the Safe Lid, at least 10% more samples can be taken per battery charge.
  • Enable tracking for digital data analysis and trending based on individual GS1 barcode.
  • Thanks to the ready-to-use and sterile format, no daily autoclaving process is required.
  • The non-twisting closure allows quick exchange of lids and supports robotic handling.
  • Individual barcodes enable tracking using digital workflows.
  • The prevention of cross-contamination during transfer into cleanrooms is ensured by the individual double packaging.
  • Sterility prevents false positives.
  • Single use followed by disposal minimizes the risk of exposure to potentially toxic airborne substances.

Legal Information

MAS-100 NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service