Skip to Content
Merck
All Photos(2)

Key Documents

1.08454

Supelco

Ethylenedinitrilotetraacetic acid disodium salt dihydrate

suitable for titration

Synonym(s):

Ethylenediaminetetraacetic acid disodium salt dihydrate, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetraacetic acid disodium salt, Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
Beilstein:
3900609
UNSPSC Code:
12352107
EC Index Number:
205-358-3
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Ethylenedinitrilotetraacetic acid disodium salt dihydrate, Ethylenedinitrilotetraacetic acid disodium salt dihydrate. CAS 6381-92-6, chemical formula C₁₀H₁₄N₂O₈*2Na*2H₂O.

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: Complexometric reactions

technique(s)

titration: suitable

pH

4-5 (20 °C, 50 g/L in H2O)

mp

248 °C (dec.) (lit.)

solubility

100 g/L

bulk density

700 kg/m3

storage temp.

no temp limit

SMILES string

O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

OVBJJZOQPCKUOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.

Analysis Note

Assay (complexometric; calculated as dihydrate): 99.0 - 101.0 %
Identity (IR-spectrum): passes test
Identity (Complexation): passes test
Identity (Sodium): passes test
pH (50 g/l; water): 4.0 - 5.5
In water insoluble matter: ≤ 0.003 %
Chloride (Cl): ≤ 0.004 %
Sulphate (SO₄): ≤ 0.01 %
Cyanide (CN): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
Cu (Copper): ≤ 0.0001 %
Fe (Iron): ≤ 0.0005 %
Pb (Lead): ≤ 0.001 %
Nitrilotriacetic acid (HPLC): ≤ 0.05 %
Loss on drying (150 °C; 6 h): 8.7 - 11.4 %

Other Notes

Elimination of water of crystallization: 110 °C

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service