Skip to Content
Merck
All Photos(1)

Key Documents

1.07810

Supelco

Tin(IV) chloride

EMPLURA®

Synonym(s):

Tin(IV) chloride, Tin tetrachloride, Stannous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl4Sn
CAS Number:
Molecular Weight:
260.52
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-588-9
form:
liquid

vapor pressure

31 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99.0% (acidimetric)

form

liquid

impurities

≤0.0005% Iodide oxidizing substances (as Cl2)
≤0.001% Iodine reducing matter (as Tin (II) chloride)
≤0.05% Substances not precipitated by hydrogen sulfide (as SO4)

pH

0.2 (20 °C, 60 g/L in H2O, Hydrolysis)

bp

114 °C/1013 hPa

mp

-33 °C

density

2.23 g/cm3 at 20 °C

cation traces

Fe: ≤0.0005%

storage temp.

2-30°C

InChI

1S/4Cl.Sn

InChI key

IPBZXRPRYYPREV-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Fe (Iron): ≤ 0.0005 %
Substances not precipitated by hydrogen sulfide (as SO₄): ≤ 0.05 %
Iodine reducing matter (as Tin (II) chloride): ≤ 0.001 %
Iodide oxidizing substances (as Cl₂): ≤ 0.0005 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service