Skip to Content
Merck
All Photos(1)

Key Documents

1.06717

Supelco

Nickel(II) chloride hexahydrate

for analysis EMSURE® ACS

Synonym(s):

Nickel(II) chloride hexahydrate, Nickel dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Ni · 6H2O
CAS Number:
Molecular Weight:
237.69
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-743-0
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
2540 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

≥98.0% (complexometric)

form

solid

potency

186 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter

pH

4.9 (20 °C, 100 g/L in H2O)

mp

140 °C (anhydrous substance; decomposes)

solubility

2540 g/L

density

1.92 g/cm3 (anhydrous substance)

bulk density

640 kg/m3

anion traces

sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Co: ≤0.005%
Cu: ≤0.001%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.01%
Mn: ≤0.002%
N: ≤0.005%
Na: ≤0.01%
Pb: ≤0.002%
Zn: ≤0.001%

storage temp.

2-30°C

SMILES string

[Ni+2].[Cl-].[Cl-].O.O.O.O.O.O

InChI

1S/2ClH.Ni.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

LAIZPRYFQUWUBN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Enhanced photocatalytic and electrochemical performance of hydrothermally prepared NiO-doped Co nanocomposites.: This study explores the synthesis of Nickel(II) chloride hexahydrate as a precursor in the hydrothermal preparation of NiO-doped Co nanocomposites. The research demonstrates significant improvements in both photocatalytic and electrochemical properties, highlighting the potential applications in advanced material science and analytical chemistry (Venkatachalapathy et al., 2024, doi:10.1002/bio.4768).

Linkage

Replaces: NX0325-5

Analysis Note

Assay (complexometric): ≥ 98.0 %
Insoluble matter: ≤ 0.005 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.005 %
Ca (Calcium): ≤ 0.005 %
Co (Cobalt): ≤ 0.005 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.01 %
Mg (Magnesium): ≤ 0.01 %
Mn (Manganese): ≤ 0.002 %
Na (Sodium): ≤ 0.01 %
Pb (Lead): ≤ 0.002 %
Zn (Zinc): ≤ 0.001 %
conforms to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service