Skip to Content
Merck
All Photos(1)

Key Documents

1.05152

Supelco

Potassium sulfate

99.999 Suprapur®

Synonym(s):

Potassium sulfate, Potassium pyrosulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K2O4S
CAS Number:
Molecular Weight:
174.26
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-915-5
NACRES:
NB.21
Assay:
99.999%
grade:
for inorganic trace analysis
form:
solid
solubility:
111 g/L
Pricing and availability is not currently available.

grade

for inorganic trace analysis

Quality Level

Assay

99.999%

form

solid

potency

6600 mg/kg LD50, oral (Rat)

pH

5.5-8.5 (20 °C, 50 g/L in H2O)

bp

1689 °C/1013 hPa

mp

1069 °C

solubility

111 g/L

density

2.66 g/cm3 at 20 °C

bulk density

800 kg/m3

storage temp.

2-30°C

SMILES string

[K+].[K+].[S](=O)(=O)([O-])[O-]

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

Related Categories

Application


  • Effect of salt modulators on the elution behavior of insulin and the separation of product-related impurities in reversed-phase chromatography.: The study examines how potassium sulfate and other salts affect the chromatography process, specifically focusing on insulin purification. The results are crucial for optimizing chromatographic methods in pharmaceutical production (Koch et al., 2023).

Analysis Note

Purity (metallic): ≥ 99.999 %
Assay (alkalimetric): ≥ 99.0
Chloride (Cl): ≤ 5 ppm
Total nitrogen (N): ≤ 5.0 ppm
Al (Aluminium): ≤ 0.050 ppm
Ba (Barium): ≤ 1.0 ppm
Bi (Bismuth): ≤ 0.01 ppm
Ca (Calcium): ≤ 0.10 ppm
Cd (Cadmium): ≤ 0.005 ppm
Ce (Cerium): ≤ 0.020 ppm
Co (Cobalt): ≤ 0.005 ppm
Cr (Chromium): ≤ 0.010 ppm
Cu (Copper): ≤ 0.020 ppm
Eu (Europium): ≤ 0.010 ppm
Fe (Iron): ≤ 0.050 ppm
Hg (Mercury): ≤ 0.05 ppm
La (Lanthanum): ≤ 0.010 ppm
Li (Lithium): ≤ 0.2 ppm
Mg (Magnesium): ≤ 0.10 ppm
Mn (Manganese): ≤ 0.020 ppm
Na (Sodium): ≤ 5.0 ppm
Ni (Nickel): ≤ 0.020 ppm
Pb (Lead): ≤ 0.050 ppm
Sc (Scandium): ≤ 0.010 ppm
Sm (Samarium): ≤ 0.010 ppm
Sr (Strontium): ≤ 0.10 ppm
Tl (Thallium): ≤ 0.01 ppm
Y (Yttrium): ≤ 0.010 ppm
Yb (Ytterbium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.050 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service