Skip to Content
Merck
All Photos(4)

Key Documents

1.01974

Supelco

n-Butyl acetate

EMPLURA®

Synonym(s):

Butyl acetate, Acetic acid n-butyl ester, Butyl ethanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)3CH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741921
MDL number:
UNSPSC Code:
12191502
EC Index Number:
204-658-1
NACRES:
NA.21
Assay:
≥99% (GC)
bp:
124-126 °C (lit.)
vapor pressure:
15 mmHg ( 25 °C)
8 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

4 (vs air)

Quality Level

vapor pressure

15 mmHg ( 25 °C)
8 mmHg ( 20 °C)

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

790 °F

potency

13100 mg/kg LD50, oral (Rat)
>14100 mg/kg LD50, skin (Rabbit)

expl. lim.

7.6 %

impurities

≤0.02% Free acid (as CH3COOH)
≤1% n-Butyl alcohol (GC)

evapn. residue

≤0.0010%

refractive index

n20/D 1.394 (lit.)

pH

6.2 (20 °C, 5.3 g/L in H2O, External MSDS)

kinematic viscosity

0.83 cSt(20 °C)

bp

124-126 °C (lit.)

mp

−78 °C (lit.)

transition temp

flash point 27 °C

density

0.88 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCOC(C)=O

InChI

1S/C6H12O2/c1-3-4-5-8-6(2)7/h3-5H2,1-2H3

InChI key

DKPFZGUDAPQIHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Exposure to Volatile Organic Compounds in Paint Production Plants: Levels and Potential Human Health Risks.: This research assesses the levels of volatile organic compounds, including n-butyl acetate, in paint production environments and discusses the associated health risks, emphasizing the need for effective exposure control strategies (Ghobakhloo et al., 2023).

  • Application of On-Line Sorbentless Cryogenic Needle Trap Extraction Coupled with GC-FID for Analysis of Some Organic Volatile Impurities in Solid Pharmaceuticals.: Research on the application of cryogenic needle trap extraction with gas chromatography for analyzing volatile impurities, including n-butyl acetate, in pharmaceutical products, demonstrating a novel approach to chemical analysis (Djozan et al., 2022).

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Free acid (as CH₃COOH): ≤ 0.02 %
Density (d 20 °C/ 4 °C): 0.879 - 0.881
Density (d 20 °C/20 °C): 0.881 - 0.883
n-Butyl alcohol (GC): ≤ 1 %
Evaporation residue: ≤ 0.0010 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service