Skip to Content
Merck
All Photos(2)

Documents

1.01773

Supelco

Petroleum benzine boiling range 40-80°C

EMPLURA®

Synonym(s):

Petroleum ether, Petroleum spirit, Petroleum ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12191502
NACRES:
NB.21

vapor pressure

400 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

form

liquid

autoignition temp.

280 °C

expl. lim.

1-7 % (v/v)

impurities

≤0.0005% Free acid (as CH3COOH)
≤0.01% Water

evapn. residue

≤0.001%

bp

40-80 °C/1013 hPa

transition temp

flash point <-30 °C

density

0.670 g/cm3 at 20 °C

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Analysis Note

Free acid (as CH₃COOH): ≤ 0.0005 %
Density (d 20 °C/ 4 °C): 0.640 - 0.670
Boiling range (40-80°C): ≥ 95 % (v/v)
Readily carbonizable substances: conforms
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.01 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-22.0 °F

Flash Point(C)

< -30 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service