1.00967
Ethanol
96%, EMPROVE® EXPERT, Ph. Eur., ChP, JP, USP
Pharma Manufacturing
Synonym(s):
Ethanol, Ethyl alcohol, EtOH, Ethyl alcohol
About This Item
Recommended Products
Agency
ChP
JP
Ph. Eur.
USP
Quality Level
vapor density
1.59 (vs air)
vapor pressure
44.6 mmHg ( 20 °C)
product line
EMPROVE® EXPERT
Assay
92.3-93.8% (d15.56°C/15.56°C; m/m)
92.6-95.2% (d20°C/4°C; m/m)
form
liquid
autoignition temp.
683 °F
expl. lim.
19 %, 60 °F
greener alternative product characteristics
Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
concentration
92.3-93.8 % (v/v) (d15°C/15°C)
95.1-96.9 % (v/v) (d15.56°C/15.56°C)
95.1-96.9 % (v/v) (d20°C/4°C)
96%
refractive index
n20/D 1.3600 (lit.)
pH
7.0 (20 °C, 10 g/L in H2O)
bp
78 °C (lit.)
mp
−114 °C (lit.)
transition temp
flash point 17 °C
density
0.805-0.812 g/cm3 at 20 °C
application(s)
liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical
semi-solid formulation
format
neat
greener alternative category
storage temp.
2-25°C
SMILES string
CCO
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3
InChI key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Due to its low microbial and endotoxin limits, Ethanol 96% Emprove® Expert is especially suitable for high-risk applications.
Footnote
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
62.6 °F
Flash Point(C)
17 °C
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Free of charge - Sign in to download
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service