Skip to Content
Merck
All Photos(1)

Documents

07-789

Sigma-Aldrich

Anti-phospho-AKT (Thr34) Antibody

Upstate®, from rabbit

Synonym(s):

Akt1, PKB, PRKBA, RAC, RAC-ALPHA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr34)

Gene Information

human ... AKT1(207)

Specificity

Predicted to cross react with Mouse and Rat due to high sequence homology, but have not been tested.
Recognizes phospho-AKT (Thr34)

Immunogen

KLH conjugated peptide corresponding to amino acid region encompassing the human phospho-AKT (Thr34); Sequence highly conserved in mouse and rat.

Application

Anti-phospho-AKT (Thr34) Antibody detects level of phospho-AKT (Thr34) & has been published & validated for use in WB.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

General Post-translation Modification

Quality

Routinely evaluated by immunoblot.

Target description

60 kDa

Physical form

100 μL of epitope specific affinity purified rabbit polyclonal IgG in 50% affinity storage buffer (PBS containing 1.0 mg/mL BSA and 0.05% sodium azide) and 50% glycerol.
ImmunoAffinity Purified

Storage and Stability

Stable for 2 years at -20°C from date of shipment. Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol-containing solutions to become frozen during storage.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cisplatin activates Akt in small cell lung cancer cells and attenuates apoptosis by survivin upregulation
Belyanskaya, Larisa L, et al
International Journal of Cancer. Journal International Du Cancer, 117, 755-763 (2005)
PKB/Akt: connecting phosphoinositide 3-kinase to cell survival and beyond.
Marte, B M and Downward, J
Trends in Biochemical Sciences, 22, 355-358 (1997)
Virginia Actis Dato et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 152, 113270-113270 (2022-06-17)
Antibodies against the P3 sequence (Gly1127-Cys1140) of LRP1 (anti-P3 Abs) specifically block cholesteryl ester (CE) accumulation in vascular cells. LRP1 is a key regulator of insulin receptor (InsR) trafficking in different cell types. The link between CE accumulation and the
Virginia Actis Dato et al.
Cells, 9(1) (2020-01-16)
Abstract: The cardiovascular disease (CVD) frequently developed during metabolic syndrome and type-2 diabetes mellitus is associated with increased levels of aggregation-prone small LDL particles. Aggregated LDL (aggLDL) internalization is mediated by low-density lipoprotein receptor-related protein-1 (LRP1) promoting intracellular cholesteryl ester
Progesterone receptor enhances breast cancer cell motility and invasion via extranuclear activation of focal adhesion kinase.
Fu, XD; Goglia, L; Sanchez, AM; Flamini, M; Giretti, MS; Tosi, V; Genazzani, AR; Simoncini, T
Endocrine-Related Cancer null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service