Skip to Content
Merck
All Photos(3)

Key Documents

07-1231

Sigma-Aldrich

Anti-Notch1 Antibody, Intracellular

serum, from rabbit

Synonym(s):

Neurogenic locus notch homolog protein 1 precursor, Notch (Drosophila) homolog 1 (translocation-associated), Notch 1, Notch homolog 1, translocation-associated (Drosophila), Translocation-associated notch protein TAN-1, Neurogenic locus notch homolog pro

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

should not react with

mouse

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NOTCH1(4851)

General description

Notch is a family of single-pass transmembrane receptor proteins that are synthesized in the endoplasmic reticulum as an inactive form and are proteolytically cleaved on an extracellular site by a furin-like convertase (S1 cleavage) in the trans-golgi network after the recognition of the RQRR sequence before it reaches the plasma membrane as heterodimers to yield an active, ligand-accessible form. The Notch family is comprised of 4 members (1-4) whose ligands include the Delta and Jagged family of ligands. These ligands cause proteolysis of Notch to liberate the intracellular domain. Cleavage results in a C-terminal fragment N(TN) and a N-terminal fragment N(EC). Following ligand binding, it is cleaved (S2 cleavage) by TNF-alpha converting enzyme (TACE) to yield a membrane-associated intermediate fragment called Notch extracellular truncation (NEXT). This fragment is then cleaved by presenilin-dependent gamma-secretase between gly1743 and val1744 (S3 cleavage) to release the intracellular domain (NICD) from the membrane. That domain translocates to the nucleus and initiates transcription events by binding the DNA binding protein CSL. The notch family members are involved cell differentiation and development.

Specificity

Notch 1, intracellular. No reactivity against human Notch 2.
Reactivity with other species has not been determined.

Immunogen

Synthetic peptide from human Notch 1.

Application

ELISA: 1:20,000-1:100,000 dilution from a previous lot was used in a standard sandwich ELISA assay against the peptide immunogen.
Optimal working dilutions must be determined by the end user.

Immunohistochemistry(paraffin):
Representative testing from a previous lot.
Optimal Staining IGF-1 With Epitope Retrieval: Lipoma.
Use Anti-Notch1 Antibody, Intracellular (Rabbit Polyclonal Antibody) validated in ELISA, WB, IHC(P) to detect Notch1 also known as Neurogenic locus notch homolog protein 1 precursor.

Quality

Routinely evaluated by Western Blot on HeLa lysates.

Western Blot Analysis: 1:500 dilution of this lot detected NOTCH1, intracellular (Cleaved form) on 10 μg of HeLa lysates.

Target description

~80 kDa

Linkage

Replaces: 04-1046

Physical form

Rabbit polyclonal IgG serum in buffer containing in 0.02 M Potassium phosphate, 0.15 M NaCl with 0.01% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Xu et al.
Andrology, 10(8), 1644-1659 (2022-09-05)
In the testis, spermatocytes and spermatids rely on lactate produced by Sertoli cells (SCs) as energy source. Transforming growth factor-beta 3 (TGF-β3) is one of the generally accepted paracrine regulatory factors of SC-created blood-testis barrier (BTB), yet its role in
LaNisha L Patterson et al.
Infection and immunity, 91(9), e0000223-e0000223 (2023-08-18)
Ehrlichia chaffeensis has evolved multiple strategies to evade innate defenses of the mononuclear phagocyte. Recently, we reported the E. chaffeensis tandem repeat protein (TRP)120 effector functions as a Notch ligand mimetic and a ubiquitin ligase that degrades the nuclear tumor
Elenaé Vázquez-Ulloa et al.
Oncotarget, 9(27), 18916-18928 (2018-05-04)
The participation of NOTCH signaling in invasive cervical cancer (ICC) remains controversial since both tumor suppressive and oncogenic properties have been described. Additionally, the role of NUMB, a negative regulator of NOTCH, remains unclear in ICC. We aimed to investigate
Xiaoyi Liu et al.
Gland surgery, 10(11), 3116-3127 (2021-12-21)
Deltex 1 (DTX1) is a single transmembrane protein with ubiquitin E3 ligase activity which has been found to play a role in the development of several cancers. We aimed to investigate the associations between DTX1 and breast cancer (BC). We
LaNisha L Patterson et al.
mBio, 13(2), e0007622-e0007622 (2022-04-01)
Ehrlichia chaffeensis evades innate host defenses by reprogramming the mononuclear phagocyte through mechanisms that involve the exploitation of multiple evolutionarily conserved cellular signaling pathways, including Notch. This immune evasion strategy is directed in part by tandem repeat protein (TRP) effectors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service