Skip to Content
Merck
All Photos(1)

Key Documents

05-525

Sigma-Aldrich

Anti-Rad50 Antibody, clone 13B3/2C6

clone 13B3/2C6, Upstate®, from mouse

Synonym(s):

Anti-NBSLD, Anti-RAD502, Anti-hRad50

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

13B3/2C6, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... RAD50(10111)
mouse ... Rad50(19360)

Specificity

Rad50

Immunogen

GST fusion protein corresponding to residues 1-425 of human Rad50

Application

Anti-Rad50 Antibody, clone 13B3/2C6 is a high quality Mouse Monoclonal Antibody for the detection of Rad50 & has been validated in IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

routinely evaluated by immunoblot on HeLa nuclear lysate

Target description

153kDa

Physical form

10mM PBS, pH 7.4
Format: Purified
Protein G Chromatography

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutations in the MRE11, RAD50, XRS2, and MRE2 genes alter chromatin configuration at meiotic DNA double-stranded break sites in premeiotic and meiotic cells.
Ohta, K, et al.
Proceedings of the National Academy of Sciences of the USA, 95, 646-651 (1998)
Functional role for senataxin, defective in ataxia oculomotor apraxia type 2, in transcriptional regulation.
Suraweera, A; Lim, Y; Woods, R; Birrell, GW; Nasim, T; Becherel, OJ; Lavin, MF
Human Molecular Genetics null
K M Trujillo et al.
The Journal of biological chemistry, 273(34), 21447-21450 (1998-08-15)
Genetic studies in yeast have indicated a role of the RAD50 and MRE11 genes in homologous recombination, telomere length maintenance, and DNA repair processes. Here, we purify from nuclear extract of Raji cells a complex consisting of human Rad50, Mre11
Bogdan I Gerashchenko et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 77(10), 940-952 (2011-02-04)
Radiosensitization of mammalian cells by heat is believed to involve the inhibition of repair of DNA double-strand breaks (DSBs). The Mre11 complex (composed of Mre11, Rad50, and Nbs1) is involved in DSB repair and forms foci at sites of radiation-induced
Tobias Hohenauer et al.
EMBO molecular medicine, 5(6), 919-934 (2013-05-15)
Pigment cells and neuronal cells both are derived from the neural crest. Here, we describe the Pit-Oct-Unc (POU) domain transcription factor Brn3a, normally involved in neuronal development, to be frequently expressed in melanoma, but not in melanocytes and nevi. RNAi-mediated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service