Skip to Content
Merck
All Photos(1)

Documents

LM2212

Avanti

C12 Ceramide

Avanti Research - A Croda Brand LM2212, ethanol solution

Synonym(s):

N-(dodecanoyl)-sphing-4-enine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H59NO3
CAS Number:
Molecular Weight:
481.79
UNSPSC Code:
12352211
NACRES:
NA.25

form

ethanol solution

packaging

pkg of 1 × 1 mL (LM2212-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM2212

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C30H59NO3/c1-3-5-7-9-11-13-14-15-16-18-19-21-23-25-29(33)28(27-32)31-30(34)26-24-22-20-17-12-10-8-6-4-2/h23,25,28-29,32-33H,3-22,24,26-27H2,1-2H3,(H,31,34)/b25-23+/t28-,29+/m0/s1

InChI key

HXFPPRPLRSPNIB-VARSQMIESA-N

General description

Ceramide is a sphingolipid with long acyl chain attached to a fatty. It is synthesized in the endoplasmic reticulum by ceramide synthase.

Biochem/physiol Actions

Ceramide functions as a secondary messenger to initiate apoptosis. It also regulates physiological processes and is accountable for the epidermal permeability barrier in eukaryotic membranes.

Packaging

2 mL Amber Glass Sealed Ampule (LM2212-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radiation and ceramide-induced apoptosis
Kolesnick R and Fuks Z
Oncogene, 22(37), 5897-5897 (2003)
The enigma of ceramide synthase regulation in mammalian cells
Wegner MS, et al.
Progress in Lipid Research, 63, 93-119 (2016)
Nesli-Ece Sen et al.
International journal of molecular sciences, 20(23) (2019-11-27)
Ataxin-2 (human gene symbol ATXN2) acts during stress responses, modulating mRNA translation and nutrient metabolism. Ataxin-2 knockout mice exhibit progressive obesity, dyslipidemia, and insulin resistance. Conversely, the progressive ATXN2 gain of function due to the fact of polyglutamine (polyQ) expansions
Mammalian Ceramide Synthases
Levy M and Futerman AH
Febs Letters, 62(5), 347-356 (2010)
Guang-Yi Dai et al.
The Journal of biological chemistry, 295(3), 717-728 (2019-12-11)
Cellular membranes contain many lipids, some of which, such as sphingolipids, have important structural and signaling functions. The common sphingolipid glucosylceramide (GlcCer) is present in plants, fungi, and animals. As a major plant sphingolipid, GlcCer is involved in the formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service