Skip to Content
Merck
All Photos(1)

Key Documents

LM1302

Avanti

17:0-20:4 PS

Avanti Research - A Croda Brand LM1302, methanol solution

Synonym(s):

1-heptadecanoyl-2-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phospho-L-serine (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H79N2O10P
CAS Number:
Molecular Weight:
815.07
UNSPSC Code:
12352211
NACRES:
NA.25

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1302-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM1302

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O.[NH4+]

InChI key

PVYYHCZONIFDMV-SPTHKXBTSA-N

General description

In humans, phosphoserine (PS) is present in the intracellular membranes and grey matter. It is localized to the nuclear envelope, endoplasmic reticulum, Golgi apparatus, outer mitochondrial membranes, plasma membranes and myelin.

Application

17:0-20:4 PS or 1-heptadecanoyl-2-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phospho-L-serine has been used as an internal standard:
  • in lipid extraction from isolated pure mitochondria for mass spectrometry (MS) analysis
  • for lipid quantification/analysis using liquid chromatography (LC)-MS
  • for phospholipid quantification by MS
  • in lysophospholipid acyltransferases (LPLAT) assay
  • in the preparation of total lipid extracts from lysosome samples by tert-butyl methyl ether (MTBE) lipid extraction protocol for analysis by LC-MS
  • for ratiometric quantification of phosphatidylserine (PS) using LC-MS

Biochem/physiol Actions

Phosphoserine (PS) maintains the nerve cell membrane and myelin sheath. It is found to be associated with cognitive disability in Alzheimer′s disease.

Packaging

2 mL Amber Glass Sealed Ampule (LM1302-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miki Eto et al.
International journal of molecular sciences, 13(12), 16267-16280 (2012-12-05)
Cellular membranes contain glycerophospholipids, which have important structural and functional roles in cells. Glycerophospholipids are first formed in the de novo pathway (Kennedy pathway) and are matured in the remodeling pathway (Lands' cycle). Recently, lysophospholipid acyltransferases functioning in Lands' cycle
Di Ding et al.
Molecular neurobiology, 56(4), 2618-2639 (2018-07-28)
Neurons from the adult central nervous system (CNS) demonstrate limited mRNA transport and localized protein synthesis versus developing neurons, correlating with lower regenerative capacity. We found that deimination (posttranslational conversion of protein-bound arginine into citrulline) undergoes upregulation during early neuronal
Penelope Dimas et al.
eLife, 8 (2019-05-08)
Oligodendrocytes (OLs) support neurons and signal transmission in the central nervous system (CNS) by enwrapping axons with myelin, a lipid-rich membrane structure. We addressed the significance of fatty acid (FA) synthesis in OLs by depleting FA synthase (FASN) from OL
Laura Montani et al.
The Journal of cell biology, 217(4), 1353-1368 (2018-02-13)
Myelination calls for a remarkable surge in cell metabolism to facilitate lipid and membrane production. Endogenous fatty acid (FA) synthesis represents a potentially critical process in myelinating glia. Using genetically modified mice, we show that Schwann cell (SC) intrinsic activity
Alexander Fauland et al.
Journal of lipid research, 52(12), 2314-2322 (2011-10-01)
This work aims to combine chromatographic retention, high mass resolution and accuracy, MS/MS spectra, and a package for automated identification and quantitation of lipid species in one platform for lipidomic analysis. The instrumental setup elaborated comprises reversed-phase HPLC coupled to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service