Skip to Content
Merck
All Photos(1)

Key Documents

870708P

Avanti

08:0 Coenzyme A

Avanti Research - A Croda Brand 870708P, powder

Synonym(s):

octanoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H59N10O17P3S
CAS Number:
Molecular Weight:
944.82
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (870708P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870708P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C29H50N7O17P3S.3H3N/c1-4-5-6-7-8-9-20(38)57-13-12-31-19(37)10-11-32-27(41)24(40)29(2,3)15-50-56(47,48)53-55(45,46)49-14-18-23(52-54(42,43)44)22(39)28(51-18)36-17-35-21-25(30)33-16-34-26(21)36;;;/h16-18,22-24,28,39-40H,4-15H2,1-3H3,(H,31,37)(H,32,41)(H,45,46)(H,47,48)(H2,30,33,34)(H2,42,43,44);3*1H3/t18-,22?,23+,24+,28-;;;/m1.../s1

InChI key

FTIQWXKUZWNHGD-DMTDIVBASA-N

General description

08:0 Coenzyme A, also referred as octanoyl Coenzyme A, is a medium-chain acyl CoA. It is a coenzyme A derivative of octanoic acid.

Application

08:0 Coenzyme A has been used as a substrate in microsomal ghrelin octanoyltransferase assay.

Biochem/physiol Actions

08:0 Coenzyme A acts as an intermediate in mitochondrial fatty acid β-oxidation. It hinders the activity of citrate synthase (CS) and glutamate dehydrogenase (GDH). Higher levels of octanoyl –CoA, due to reduced β-oxidation of octanoyl-CoA by medium-chain acyl CoA dehydrogenase (MCADH), is observed in patients with MCADH deficiency (MCD).

Packaging

5 mL Amber Glass Screw Cap Vial (870708P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Modre-Osprian et al.
BMC systems biology, 3, 2-2 (2009-01-08)
The oxidation of fatty acids in mitochondria plays an important role in energy metabolism and genetic disorders of this pathway may cause metabolic diseases. Enzyme deficiencies can block the metabolism at defined reactions in the mitochondrion and lead to accumulation
B A Amendt et al.
The Journal of clinical investigation, 76(3), 963-969 (1985-09-01)
Medium-chain acyl-coenzyme A (CoA) dehydrogenase (MCADH; EC 1.3.99.3) deficiency (MCD) is an inborn error of beta-oxidation. We measured 3H2O formed by the dehydrogenation of [2,3-3H]acyl-CoAs in a 3H-release assay. Short-chain acyl-CoA dehydrogenase (SCADH; EC 1.3.99.2), MCADH, and isovaleryl-CoA dehydrogenase (IVDH;
J C Lai et al.
Research communications in chemical pathology and pharmacology, 82(3), 331-338 (1993-12-01)
We investigated the hypothesis that one mechanism underlying fatty acid toxicity is the selective inhibition of rate-limiting and/or regulated tricarboxylic acid cycle and related enzymes by fatty acyl coenzyme A (CoA) derivatives by examining the effects of several fatty acyl
Martin S Taylor et al.
Bioorganic chemistry, 62, 64-73 (2015-08-08)
Ghrelin-O-Acyltransferase (GOAT) is an 11-transmembrane integral membrane protein that octanoylates the metabolism-regulating peptide hormone ghrelin at Ser3 and may represent an attractive target for the treatment of type II diabetes and the metabolic syndrome. Protein octanoylation is unique to ghrelin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service