Skip to Content
Merck
All Photos(1)

Key Documents

870430O

Avanti

C20:4 anandamide (AEA)

Avanti Research - A Croda Brand 870430O

Synonym(s):

5Z,8Z,11Z,14Z-eicosatetraenoylethanolamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H37NO2
CAS Number:
Molecular Weight:
347.53
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 5 mg (870430O-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870430O

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)NCCO

InChI

1S/C22H37NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-22(25)23-20-21-24/h6-7,9-10,12-13,15-16,24H,2-5,8,11,14,17-21H2,1H3,(H,23,25)/b7-6-,10-9-,13-12-,16-15-

InChI key

LGEQQWMQCRIYKG-DOFZRALJSA-N

General description

Anandamide (AEA) is a lipid neurotransmitter, which is produced and released by the neurons in the brain and macrophages. It is an N-acyl ethanolamide derivative of C20,4 fatty acid. AEA is an endogenous ligand for cannabinoid receptors.

Biochem/physiol Actions

Anandamide (AEA) released by the macrophages exhibits anti-inflammatory effect on the central and peripheral nervous system.

Packaging

5 mL Amber Glass Screw Cap Vial (870430O-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anandamide and its metabolites: what are their roles in the kidney?
Ritter J K, et al.
Frontiers in Bioscience, 8(11), 264-264 (2016)
Anandamide receptors
Di Marzo V, et al.
Prostaglandins, Leukotrienes, and Essential Fatty Acids, 66(2-3), 377-391 (2002)
Targeting fatty-acid amide hydrolase with prodrugs for CNS-selective therapy
Meinig J M, et al.
ACS Chemical Neuroscience, 8(11), 2468-2476 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service