Skip to Content
Merck
All Photos(1)

Key Documents

860668P

Avanti

4E,11Z-Sphingadiene

Avanti Research - A Croda Brand 860668P, powder

Synonym(s):

(2S,3R,4E,11Z)-2-aminooctadec-4,11-diene-1,3-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H35NO2
CAS Number:
Molecular Weight:
297.48
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860668P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860668P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](N)([H])[C@]([H])(O)/C=C/CCCCC/C=C\CCCCCC

InChI

1S/C18H35NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h7-8,14-15,17-18,20-21H,2-6,9-13,16,19H2,1H3/b8-7-,15-14+/t17-,18+/m0/s1

InChI key

PZYQAHLJANGLKE-ZLHPJBHGSA-N

General description

4E,11Z-Sphingadiene is a unique sphingoid base containing trans double bond at C4 and cis double bond at C11 positions along the sphingoid base alkyl chain. Sphingadienes are found in insects, marine organisms, legumes including soybeans and soy products.

Application

4E,11Z-Sphingadiene has been used as a standard in 193 nm ultraviolet photodissociation (UVPD)-ultra high resolution tandem mass spectrometry for its detailed structural characterization.

Biochem/physiol Actions

Sphingadienes are growth-inhibitory sphingoid bases, involved in apoptosis and cell signaling. It inhibits Akt/Wnt signaling that might contribute to their chemopreventive properties in colon cancer.

Packaging

5 mL Amber Glass Screw Cap Vial (860668P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detailed structural characterization of sphingolipids via 193 nm ultraviolet photodissociation and ultrahigh resolution tandem mass spectrometry
Ryan E, et al.
Journal of the American Society For Mass Spectrometry, 28(7), 1406-1419 (2017)
Ashok Kumar et al.
Carcinogenesis, 33(9), 1726-1735 (2012-05-15)
Sphingadienes (SDs) derived from soy and other natural sphingolipids are cytotoxic to colon cancer cells via an Akt-dependent mechanism and reduce adenoma formation in Apc(Min/+) mice. Wnt signaling is fundamental to colon carcinogenesis and is the basis for spontaneous tumorigenesis
Occurrence, structure elucidation, biosynthesis, functions and synthesis of sphingadienes
U Abeytunga T
Mini-Reviews in Organic Chemistry, 12(3), 282-292 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service