Skip to Content
Merck
All Photos(1)

Documents

860545P

Avanti

C12 Lactosyl(β) Ceramide (d18:1/12:0)

Avanti Research - A Croda Brand 860545P, powder

Synonym(s):

D-lactosyl-β-1,1′ N-lauroyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H79NO13
CAS Number:
Molecular Weight:
806.08
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (860545P-25mg)
pkg of 1 × 5 mg (860545P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860545P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O[C@H](O2)[C@H](O)[C@@H](O)[C@H]([C@H]2CO)O

InChI

1S/C42H79NO13/c1-3-5-7-9-11-13-14-15-16-18-19-21-23-25-31(46)30(43-34(47)26-24-22-20-17-12-10-8-6-4-2)29-53-41-39(52)37(50)40(33(28-45)55-41)56-42-38(51)36(49)35(48)32(27-44)54-42/h23,25,30-33,35-42,44-46,48-52H,3-22,24,26-29H2,1-2H3,(H,43,47)/b25-23+/t30-,31+,32?,33?,35-,36?,37?,38?,39?,40+,41+,42-/m0/s1

InChI key

KNWHKVBHCLQVFX-NKNVVQMUSA-N

Application

C12 Lactosyl(β) Ceramide (d18:1/12:0) or D-lactosyl-β-1,1′ N-lauroyl-D-erythro-sphingosine may be used:
  • as a lipid standard for the quantification of lipid in planarians using mass spectrometry
  • in lipid binding assay to assess the binding of monoclonal antibodies (mAbs)
  • as an internal standard for lipid extraction from mice using Bligh and Dyer procedure

Packaging

5 mL Amber Glass Screw Cap Vial (860545P-25mg)
5 mL Amber Glass Screw Cap Vial (860545P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mithilesh Kumar Jha et al.
Glia, 68(1), 161-177 (2019-08-28)
Schwann cell (SC)-specific monocarboxylate transporter 1 (MCT1) knockout mice were generated by mating MCT1 f/f mice with myelin protein zero (P0)-Cre mice. P0-Cre+/- , MCT1 f/f mice have no detectable early developmental defects, but develop hypomyelination and reduced conduction velocity
Albert Thommen et al.
eLife, 8 (2019-01-05)
Kleiber's law, or the 3/4 -power law scaling of the metabolic rate with body mass, is considered one of the few quantitative laws in biology, yet its physiological basis remains unknown. Here, we report Kleiber's law scaling in the planarian
Shelly J Krebs et al.
Immunity, 50(3), 677-691 (2019-03-17)
Lineage-based vaccine design is an attractive approach for eliciting broadly neutralizing antibodies (bNAbs) against HIV-1. However, most bNAb lineages studied to date have features indicative of unusual recombination and/or development. From an individual in the prospective RV217 cohort, we identified
David B Iaea et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1865(4), 158609-158609 (2020-01-10)
STARD4, a member of the evolutionarily conserved START gene family, is a soluble sterol transport protein implicated in cholesterol sensing and maintenance of cellular homeostasis. STARD4 is widely expressed and has been shown to transfer sterol between liposomes as well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service