Skip to Content
Merck
All Photos(1)

Key Documents

860086P

Avanti

Ganglioside GQ1b

powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C106H194N10O55
Molecular Weight:
2488.71
UNSPSC Code:
12352211
NACRES:
NA.28

Assay

99% (TLC)

form

powder

packaging

package of 1 × 1 mg (860086-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids
natural lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCCC)=O)CO[C@H]1[C@H](O)[C@@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@@H]([C@H](O[C@@]3(C([O-])=O)O[C@@H]([C@H](O)[C@H](O[C@@]4(C([O-])=O)O[C@@H]([C@H](O)[C@H](O)CO)[C@H](NC(C)=O)[C@@H](O)C4)CO)[C@@](NC(C)=O)

General description

Ganglioside GQ1b is a ganglioside, which contains tetra-sialosyl group and a disialosyl group attached to the internal galactose. Ganglioside is a glycosphingolipid and is localized in the cell membrane and mammalian brain. It is attached to the external leaflet of the lipid bilayer via a ceramide moiety.

Biochem/physiol Actions

Gangliosides are involved in several neurophysiological functions, such as synaptogenesis, synaptic transmission, neurogenesis and proliferation.

Packaging

5 mL Amber Glass Screw Cap Vial (860086P-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillain-Barre syndrome and anti-ganglioside antibodies: a clinician-scientist?s journey
Yuki N.
Proceedings of the Japan Academy. Series B, Physical and Biological Sciences, 88(7), 299-326 (2012)
Ganglioside GQ1b improves spatial learning and memory of rats as measured by the Y-maze and the Morris water maze tests
Jung W R, et al.
Neuroscience Letters, 439(2), 220-225 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service