Skip to Content
Merck
All Photos(2)

Key Documents

857120P

Avanti

14:0 Lyso PA

1-myristoyl-2-hydroxy-sn-glycero-3-phosphate (sodium salt), powder

Synonym(s):

1-tetradecanoyl-sn-glycero-3-phosphate (sodium salt); PA(14:0/0:0); 14:0 LPA; 110677

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H34O7PNa
CAS Number:
Molecular Weight:
404.41
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPA; may contain up to 10% of the 2-LPA isomer, TLC)

form

powder

packaging

pkg of 1 × 25 mg (857120P-25mg)
pkg of 1 × 500 mg (857120P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857120P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(O)=O)([H])COC(CCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C17H35O7P.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-17(19)23-14-16(18)15-24-25(20,21)22;/h16,18H,2-15H2,1H3,(H2,20,21,22);/q;+1/p-1/t16-;/m1./s1

InChI key

HGENTPHNTSLRPB-PKLMIRHRSA-M

General description

Lysophosphatidic acid (LPA) belongs to a class of phospholipids, that has a glycerol backbone attached by a phosphate group, an aliphatic chain and a hydroxyl group. It has a phosphate group attached to sn-3 position, an aliphatic chain attached to sn-1 or sn-2 position and the hydroxyl group attached to the remaining sn-1 or sn-2 position.

Application

14:0 Lyso PA is suitable to use as internal standard to spike serum samples for the extraction of serum lipids. It is also suitable to use as standard lipids for extraction of lipids and for quality control in multiple reaction monitoring (MRM) assay.

Biochem/physiol Actions

Lysophosphatidic acid (LPA) participates in several biological functions. It acts as a potential biomarker for detecting ovarian cancer. LPA stimulates platelet aggregation, smooth muscle contraction, chemotaxis, cytoskeleton rearrangement, Ca2+ mobilization, neurotransmitter release and cell proliferation.

Packaging

20 mL Clear Glass Screw Cap Vial (857120P-500mg)
5 mL Amber Glass Screw Cap Vial (857120P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultra-high-performance liquid chromatography electrospray ionization tandem mass spectrometry for accurate analysis of glycerophospholipids and sphingolipids in drug resistance tumor cells
Li L, et al.
Journal of Chromatography A, 1381(2), 140-148 (2015)
Determination of lysophosphatidic acids by capillary electrophoresis with indirect ultraviolet detection
Chen YL and Xu Y
Journal of Chromatography. B, Biomedical Sciences and Applications, 753(2), 355-363 (2001)
Quantitative determination of lysophosphatidic acid by LC/ESI/MS/MS employing a reversed phase HPLC column
Shan L, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 864(1-2), 22-28 (2008)
Olaposi I Omotuyi et al.
Scientific reports, 5, 13343-13343 (2015-08-14)
Lysophosphatidic acid (LPA) receptor 1 (LPA1) is a member of the G protein-coupled receptors mediating the biological response to LPA species. Lack of detailed mechanism underlying LPA/LPA1 interaction has hampered the development of specific antagonists. Here, novel N-terminal Lys39 has
Jesica R Williams et al.
The Journal of biological chemistry, 284(25), 17304-17319 (2009-04-16)
Lysophosphatidic acid (LPA) is a ligand for LPA(1-3) of the endothelial differentiation gene family G-protein-coupled receptors, and LPA(4-8) is related to the purinergic family G-protein-coupled receptor. Because the structure-activity relationship (SAR) of GPR92/LPA(5) is limited and whether LPA is its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service