Skip to Content
Merck
All Photos(2)

Documents

852446C

Avanti

16:0 dimethylacetal

hexadecanal-dimethylacetal, chloroform

Synonym(s):

1,1-Dimethoxyhexadecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H38O2
CAS Number:
Molecular Weight:
286.49
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (852446C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 852446C

concentration

10 mg/mL (852446C-10mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(OC)OC

InChI

1S/C18H38O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19-2)20-3/h18H,4-17H2,1-3H3

InChI key

LACZJJJYINHZIR-UHFFFAOYSA-N

General description

DMAs are an indirect marker of levels of plasmalogens in plasma and may be used as standards for quantitation of plasmalogen in biological samples.

Biochem/physiol Actions

Dimethylacetals functions as an indirect marker of levels of plasmalogens, endogenous antioxidants. It plays a major part in oxidative protection and cholesterol homeostasis. Dimethylacetal may be used as standards for quantitation of plasmalogen in biological samples.

Packaging

5 mL Clear Glass Sealed Ampule (852446C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimethyl acetals, an indirect marker of the endogenous antioxidant plasmalogen level, are reduced in blood lipids of Sudanese pre-eclamptic subjects whose background diet is high in carbohydrate
Bueno AA, et al.
The Journal of Obstetrics and Gynecology of India, 32(3), 241-246 (2012)
Marina Moraitou et al.
Blood cells, molecules & diseases, 41(2), 196-199 (2008-05-27)
Plasmalogens represent a unique type of phospholipids characterized by the presence of a vinyl-ether bond at the sn-1 position of the glycerol backbone. Peroxisomes are essential in their biosynthesis. Their suggested functions include protection against oxidative stress, participation in signal
T Brosche
Archives of gerontology and geriatrics, 32(3), 283-294 (2001-06-08)
The precise role played by plasmalogen phospholipids (PL) of lipoproteins and cell membranes is not well understood. However, they might act as endogenous antioxidants in defending cell membranes and lipoproteins from reactive oxygen species. A decline of plasmalogen concentrations has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service