Skip to Content
Merck
All Photos(2)

Key Documents

840500C

Avanti

16:0-22:6 PG

1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform

Synonym(s):

1-hexadecanoyl-2-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); PG(16:0/22:6(4Z,7Z,10Z,13Z,16Z,19Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H74O10PNa
CAS Number:
Molecular Weight:
817.02
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840500C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840500C

concentration

10 mg/mL (840500C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CC)COC(=O)CCCCCCCCCCCCCCC)OCC(O)CO

InChI

1S/C44H75O10P.Na/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-24-26-28-30-32-34-36-44(48)54-42(40-53-55(49,50)52-38-41(46)37-45)39-51-43(47)35-33-31-29-27-25-23-16-14-12-10-8-6-4-2;/h5,7,11,13,17-18,20-21,24,26,30,32,41-42,45-46H,3-4,6,8-10,12,14-16,19,22-23,25,27-29,31,33-40H2,1-2H3,(H,49,50);/q;+1/p-1/b7-5-,13-11-,18-17-,21-20-,26-24-,32-30-;/t41?,42-;/m1./s1

InChI key

PPLIUYJYDZUMAR-DDKPFXSLSA-M

General description

16:0-22:6 PG or 1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phospho-(1′-rac-glycerol) is a sodium salt of glycerophospholipid, with substitution of palmitic acid, docosahexaenic acid and phospho-rac-(1-glycerol) at the sn-1, sn-2 and sn-3 positions respectively, in the glycerol backbone. Phosphatidylglycerol (PG) is an anionic lipid, enriched in several bacterial membranes. It is present in animal tissues, mainly in the lung surfactant.

Biochem/physiol Actions

Phosphatidylglycerol (PG) is useful in preparing nanoliposomes.

Packaging

5 mL Clear Glass Sealed Ampule (840500C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of complex lipidomes
Medical Applications of Mass Spectrometry, 223-249 (2008)
Pulmonary Surfactant Trafficking and Homeostasis
Lung Epithelial Biology in the Pathogenesis of Pulmonary Disease, 59-75 (2017)
Nanoparticles: A novel approach to target tumors
Nano- and Microscale Drug Delivery Systems, 113-129 (2017)
Analysis of complex lipidomes
Medical Applications of Mass Spectrometry, 223-249 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service