Skip to Content
Merck
All Photos(1)

Key Documents

700054P

Avanti

5α,6β-dihydroxycholestanol

Avanti Research - A Croda Brand

Synonym(s):

3β,5α,6β-trihydroxycholestane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H48O3
CAS Number:
Molecular Weight:
420.67
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

description

Cholestane-3β,5α,6β-triol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (700054P-25mg)
pkg of 1 × 5 mg (700054P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@]21[C@@]([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@H](CCCC(C)C)C)(CC3)C)C[C@H]2O)(CC[C@@H](C1)O)C

InChI

1S/C27H48O3/c1-17(2)7-6-8-18(3)21-9-10-22-20-15-24(29)27(30)16-19(28)11-14-26(27,5)23(20)12-13-25(21,22)4/h17-24,28-30H,6-16H2,1-5H3/t18-,19+,20+,21-,22+,23+,24-,25-,26-,27+/m1/s1

InChI key

YMMFNKXZULYSOQ-RUXQDQFYSA-N

General description

5α,6β-dihydroxycholestanol is a structural derivative of cholesterol. It is produced by the low-density lipoprotein oxidation (LDL) and is a cholesterol autooxidation product. 5α,6β-dihydroxycholestanol is an abundant oxysterol.

Application

5α,6β-dihydroxycholestanol has been used as a cholesterol oxidation product in reversed phase chromatography (RP-HPLC) and mass spectroscopy analysis. It may be used as an internal standard in tandem mass spectrometry (MS/MS analysis) of plasma samples.

Biochem/physiol Actions

5α,6β-dihydroxycholestanol (5α,6α-diOH-Chol) or cholestan-3β,5α,6β-triol (C-triol) levels are elevated along with 7-ketocholesterol in Niemann-Pick type C (NP-C) disease and is a key biomarkers for detection. It exhibits neuroprotective functionality during central nervous system (CNS) ischemia.

Packaging

5 mL Amber Glass Screw Cap Vial (700054P-25mg)
5 mL Amber Glass Screw Cap Vial (700054P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ren Sheng et al.
Nature communications, 5, 4393-4393 (2014-07-16)
Wnt proteins control diverse biological processes through β-catenin-dependent canonical signalling and β-catenin-independent non-canonical signalling. The mechanisms by which these signalling pathways are differentially triggered and controlled are not fully understood. Dishevelled (Dvl) is a scaffold protein that serves as the
Sara Boenzi et al.
Journal of lipid research, 57(3), 361-367 (2016-01-07)
Oxysterols are intermediates of cholesterol metabolism and are generated from cholesterol via either enzymatic or nonenzymatic pathways under oxidative stress conditions. Cholestan-3β,5α,6β-triol (C-triol) and 7-ketocholesterol (7-KC) have been proposed as new biomarkers for the diagnosis of Niemann-Pick type C (NP-C)
Haiyan Hu et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(34), 11426-11438 (2014-08-22)
Overstimulation of NMDA-type glutamate receptors is believed to be responsible for neuronal death of the CNS in various disorders, including cerebral and spinal cord ischemia. However, the intrinsic and physiological mechanisms of modulation of these receptors are essentially unknown. Here
G J Schroepfer
Physiological reviews, 80(1), 361-554 (2000-01-05)
Oxygenated derivatives of cholesterol (oxysterols) present a remarkably diverse profile of biological activities, including effects on sphingolipid metabolism, platelet aggregation, apoptosis, and protein prenylation. The most notable oxysterol activities center around the regulation of cholesterol homeostasis, which appears to be
Changguang Wang et al.
International journal of pharmaceutics, 569, 118576-118576 (2019-07-31)
Cholesterol is one of the major structural constituents in a liposomal bilayer. Cholesterol is susceptible to various reactions in the presence of oxygen, heat, light, certain metals, and radicals during manufacturing or storage, which may cause to generate cholesterol oxidation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service