Skip to Content
Merck
All Photos(2)

Key Documents

700027P

Avanti

7α-hydroxy-3-oxo-4-cholestenoic acid

Avanti Research - A Croda Brand

Synonym(s):

(25R)-cholest-4-en-26-oic acid, 7α-hydroxy-3-oxo

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H42O4
CAS Number:
Molecular Weight:
430.62
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700027P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

7α-Hydroxy-3-oxo-4-cholestenoic acid (7-HOCA) is synthesized extrahepatically from cholesterol. The enzyme oxysterols-7α-hydrolase (CYP7B1) acts on precursor 27-hydroxycholesterol (27-OHC) to generate 7-HOCA.. 7-HOCA is catabolized in the liver.

Application

7α-hydroxy-3-oxo-4-cholestenoic acid has been used as an analyte in ultra-high-performance liquid chromatography (UHPLC) system.

Biochem/physiol Actions

The levels of 7α-hydroxy-3-oxo-4-cholestenoic acid (7-HOCA) in cerebrospinal fluid are not correlated to neurodegeneration but may serve as a diagnostic marker to trace blood-brain barrier integrity.

Packaging

5 mL Amber Glass Screw Cap Vial (700027P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marked accumulation of 27-hydroxycholesterol in the brains of Alzheimer's patients with the Swedish APP 670/671 mutation
Shafaati M, et al.
Journal of Lipid Research, 52(5), 1004-1010 (2011)
Cholestenoic acid is a prognostic biomarker in acute respiratory distress syndrome
Madenspacher JH, et al.
The Journal of Allergy and Clinical Immunology, 143(1), 440-442 (2019)
7 ? -hydroxy-3-oxo-4-cholestenoic acid in cerebrospinal fluid reflects the integrity of the blood-brain barrier
Saeed A, et al.
Journal of Lipid Research, 55(2), 313-318 (2014)
Changes in brain oxysterols at different stages of Alzheimer's disease: Their involvement in neuroinflammation
Testa G, et al.
Redox Biology, 10, 24-33 (2016)
Vladimiro Cardenia et al.
Biochimie, 153, 99-108 (2018-08-06)
The electronic cigarettes (e-cigarettes, e-cigs) have become the most sought-after alternative to the traditional cigarettes, partly due to the widespread perception of safety. However, the high temperature reached by e-cig solutions can generate toxic compounds, some of which are listed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service