Key Documents
Z414166
Sigma-Aldrich
Aldrich® dual bank manifold with high-vacuum PTFE valves
standard manifold, positions, 5
About This Item
UNSPSC Code:
40161501
NACRES:
NB.79
Pricing and availability is not currently available.
feature
positions 5
L
500 mm
Related Categories
General description
The dual bank manifolds come with 0-10 mm bore high-vacuum PTFE valves.
Attention: We supply high quality PTFE valves on Aldrich brand glassware; these have been tested and approved by our Glass Center.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)