Skip to Content
Merck
All Photos(1)

Key Documents

Z281778

Sigma-Aldrich

Chem-FLEX transfer line

tube L 120 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.43
Pricing and availability is not currently available.

tube L

120 in.

Looking for similar products? Visit Product Comparison Guide

General description

Two 12 gauge SS needles (6 and 18 in.) are connected to the CHEM-FLEX 106 tubing with nylon clamps. Liquids contact only PTFE and SS during transfers. For extra security when using large bore transfer needles, fasten septum to bottle or flask opening with a wrap-it tie.

The nylon clamps are replaceable, see Z224170.

Please note: To see all 58 stainless steel 304 syringe needles with non-coring points click here

Legal Information

Chem-Flex is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service